Skip to Content
Merck

Skip To

L2630

Lipopolysaccharides from Escherichia coli O111:B4

purified by phenol extraction

Lipopolysaccharides from Escherichia coli O111:B4

Synonym(s):

LPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
EC Number:
297-473-0
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli (O111:B4)

Quality Segment

form

lyophilized powder

purified by

phenol extraction

technique(s)

ELISA: suitable, cell based assay: suitable, ligand binding assay: suitable

impurities

≤3.00% protein (Lowry-TCA)

color

off-white to faint yellow

solubility

water: soluble

application(s)

cell analysis
clinical research
life science and biopharma

shipped in

ambient

storage temp.

2-8°C

General description

Lipopolysaccharide is a glycolipid, that is composed of carbohydrates linked to fatty acids.[1] Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. A typical LPS molecule has lipid A (a glucosamine-based phospholipid), a relatively short core oligosaccharide and a distal polysaccharide (O-antigen). LPSs contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics. They stimulate host cells via TLR2 and TLR4 (toll like receptors). This stimulation results in the generation of various proinflammatory cytokines, such as TNFα (tumor necrosis factor), IL-1 (interleukin 1) and IL-6.[2]
This product is phenol extracted from Escherichia coli serotype O111:B4. The source strain is from a private collection. This LPS serotype has been used to stimulate B-cells and induce NOS (nitric oxide synthase) in human hepatocytes.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).
Lipopolysaccharides (LPSs) from Escherichia coli O111:B4 have been used:

  • for the stimulation of macrophages
  • for LPS preconditioning in mice
  • to induce acute kidney injury in mice
  • to induce acute respiratory distress syndrome (ARDS) in mice to study the anti-inflammatory effects of bone marrow mesenchymal stem cell (BMSCs)-derived exosomes[3]
  • to induce hyperinflammatory status in severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infected mice

Biochem/physiol Actions

Lipopolysaccharides (LPS) serve as a part of the outer membrane, this makes the outer membrane permeable to hydrophobic molecules such as antimicrobial substances that make Gram-negative bacteria innately resistant. LPS functions in bacteria-host interactions that regulate the host system responses.[1]

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L2880L3129L3012
biological source

Escherichia coli (O111:B4)

biological source

Escherichia coli (O55:B5)

biological source

Escherichia coli (O127:B8)

biological source

Escherichia coli (O111:B4)

technique(s)

ELISA: suitable, ligand binding assay: suitable, cell based assay: suitable

technique(s)

cell based assay: suitable

technique(s)

-

technique(s)

-

solubility

water: soluble

solubility

water: soluble

solubility

water: soluble

solubility

water: soluble

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

300

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 15 Questions  
  1. Is this product considered a rough strain or smooth strain?

    1 answer
    1. This product (lipopolysaccharide from E. coli O111:B4) is classified as a smooth (S-type) lipopolysaccharide. Please see the link below to review a publication that may be helpful:
      https://www.ncbi.nlm.nih.gov/books/NBK554414/

      Helpful?

  2. Can I resuspend this LPS in PBS instead of water, without hindering its performance ?

    1 answer
    1. This product can be resuspended in PBS instead of water. Both water and PBS are suitable solvents, and a hazy solution will form in either.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  4. When I compared COAs of two batches of LPS (L2630) we have been using, I realized the potency of the newer one is 10-fold higher than the older one (30000000 and 3000000 EU/mg). Is it common for LPS that different lots have such different potency?

    1 answer
    1. Significant lot-to-lot variability in potency is quite normal for this product. It is a somewhat crude phenol extract; it is not chromatographically purified. That is, a chromatographically purified LPS such as Product L3012 would be expected to have better lot-to-lot consistency. Note that for both this product and L3012, the potency specification is greater than or equal to 500,000 Endotoxin units per milligram (EU/mg); there is a lower limit, but there is no upper limit.

      Helpful?

  5. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  6. Is there any harm in filter sterilizing a solution with this LPS before using it in cell culture or animals? Meaning does it change or lose potency if it is sterile filtered say through a 0.2 micron filter?

    1 answer
    1. Sterile filtration should not affect the concentration or properties of the solution if the LPS is fully dissolved, but please keep in mind that this is a research-grade material that has not been best tested on animals. For a cell culture grade material that has already been γ-irradiated, please consider the following: https://www.sigmaaldrich.com/product/sigma/l4391

      Helpful?

  7. I received LPS L2630 at Room Temperature. Is it ok to keep it at Room Temperature before reconstitution, although recommendations are to store it between 2-8 C?

    1 answer
    1. This product ships routinely at ambient temperature and is stable up to 2 weeks to allow for transit time. The item is to be stored at 2-8°C when received.

      Please see the link below to review the product storage and shipping guidelines:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/232/426/storage-transport-conditions-ms.pdf

      Helpful?

  8. Is product L2630 suitable for use in mammalian cell culture?

    1 answer
    1. Specific testing or validation for the use of product L2630 in mammalian cell culture has not been conducted.

      Helpful?

  9. Hi i recieved L2630 10mg Lipopolysaccharides from Escherichia coli O111:B4 i will used in experimental work. how i will dilute it

    1 answer
    1. The product is soluble in water (5 mg/mL) or cell culture medium (1 mg/mL). Please see the link below to review additional preparation and storage information:
      https://www.sigmaaldrich.com/US/en/technical-documents/protocol/clinical-testing-and-diagnostics-manufacturing/bacteriology/lipopolysaccharides

      Helpful?

  10. What is the purity level of this product ? is it ultrapure? 100% pure LPS ?

    1 answer
    1. This product is not tested for purity. However, it is tested for potency on a lot-specific basis.

      Kindly review a sample CoA accessible at this link: https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/L2630-VAR0000271356.pdf

      Helpful?

1–10 of 15 Questions  

Reviews

Active Filters

  1. 1 Ratings-only review