Skip to Content
Merck
All Photos(1)

Key Documents

SMB00076

Sigma-Aldrich

Prunin

≥95% (LC/MS-ELSD)

Synonym(s):

Naringenin-7-O-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22O10
CAS Number:
Molecular Weight:
434.39
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2cc3c(c(c2)O)C(=O)C[C@H](O3)c4ccc(cc4)O

InChI

1S/C21H22O10/c22-8-16-18(26)19(27)20(28)21(31-16)29-11-5-12(24)17-13(25)7-14(30-15(17)6-11)9-1-3-10(23)4-2-9/h1-6,14,16,18-24,26-28H,7-8H2/t14-,16+,18+,19-,20+,21+/m0/s1

InChI key

DLIKSSGEMUFQOK-SFTVRKLSSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Choi et al.
Planta medica, 57(3), 208-211 (1991-06-01)
Blood glucose and total lipid levels in rats with streptozotocin-induced diabetes were determined after intraperitoneal administration of a methanolic extract of Prunus davidiana Fr. stems and its main component, prunin (= naringenin 7-O-beta-D-glucoside). From the data obtained it was concluded
LeAnna N Willison et al.
Molecular immunology, 55(3-4), 253-263 (2013-03-19)
Tree nuts are a widely consumed food. Although enjoyed safely by most individuals, allergic reactions to tree nuts, including almond, are not uncommon. Almond prunin (Pru du 6), an 11S globulin (legumin), is an abundant nut seed protein and a
K Habelt et al.
Analytical biochemistry, 134(2), 393-397 (1983-10-15)
A naringinase assay capable of distinguishing between the content of naringin, prunin, and naringenin present in the incubation mixture, is described. The amount of these compounds can be estimated by combining two spectrophotometric procedures. (a) Treatment with strong alkali to
J Garcia-Mas et al.
Plant molecular biology, 27(1), 205-210 (1995-01-01)
A number of different cDNA clones corresponding to the most abundant mRNAs present in immature seeds have been isolated from an almond (Prunus amygdalus cv. Texas) immature seed cDNA library. Those corresponding to proteins involved in storage processes have been
Hasim Kelebek et al.
Food chemistry, 220, 31-41 (2016-11-20)
The effects of hot and cold break industrial tomato paste production steps on phenolic compounds, carotenoids, organic acids, hydroxy methyl furfural (HMF) and other quality parameters of tomato pastes were investigated in this study. Phenolic compounds, carotenoids, organic acids, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service