Skip to Content
Merck
All Photos(1)

Key Documents

MWGF1000

Sigma-Aldrich

Gel Filtration Markers Kit for Protein Molecular Weights 29,000-700,000 Da

Synonym(s):

Gel Filtration Markers Kit, Gel Filtration Protein Ladder, Protein Molecular Weight Marker Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

form

liquid

technique(s)

liquid chromatography (LC): suitable

storage temp.

−20°C

General description

Gel filtration chromatography is a well-known technique for measuring the size and molecular weight of proteins. It separates proteins based on their ability to diffuse into the pores of the resin. Larger proteins have more difficulty entering the resin pores, so they flow through the column more quickly than smaller proteins. As a result, the proteins are eluted from the column in order of decreasing molecular weight.

Application

Gel Filtration Markers Kit for Protein Molecular Weights 29,000-700,000 Da has been used:
  • for protein purification and analyses (gel-filtration experiments)
  • for size-exclusion chromatography/gel filtration chromatography of synaptoneurosome extract
  • for glycerol gradient centrifugation
  • to study glucocorticoid-induced lymphocytolysis as a model system for apoptosis within the immune system
  • to isolate Polygonatum odoratum lectin, which showed remarkable anti-HSV-II activity towards Vero cells, cytotoxicity towards human melanoma A375 cells and induced apoptosis in a caspase-dependent manner

Biochem/physiol Actions

The method described in the bulletin for determining molecular masses using gel filtration chromatography is a modified version of existing published techniques. The protein standards included in this kit may be compatible with other chromatographic systems like high-performance liquid chromatography (HPLC). However, certain buffer systems may affect the elution volumes of albumin and carbonic anhydrase. The proteins in this kit have a molecular mass range spanning from 29 kDa to 699 kDa.

Other Notes

  • For research use only. Not for drug, household, or other uses.
  • Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
  • Please refer to the product information sheet to learn more about reagents, storage and stability, and procedure.

Kit Components Also Available Separately

Product No.
Description
SDS

  • A8531Albumin, bovine serum, 66,000 MW 1 mL/vialSDS

  • A8656Alcohol Dehydrogenase, yeast, 150,000 MW 1 mL/vialSDS

  • A8781 β-Amylase, sweet potato, 200,000 MW 1 mL/vialSDS

  • A3660 Apoferritin, horse spleen, 443,000 MW 1 mL/vialSDS

  • D4772Blue dextran, 2,000,000 MW 1 mL/vialSDS

  • C7025Carbonic Anhydrase, bovine erythrocytes, 29,000 MW 1 mL/vialSDS

  • T9145Thyroglobulin, bovine, 669,000 MW 1 mL/vialSDS

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Yue et al.
Nucleic acids research, 48(17), 9589-9605 (2020-08-29)
Transcription termination defines accurate transcript 3'-ends and ensures programmed transcriptomes, making it critical to life. However, transcription termination mechanisms remain largely unknown in Archaea. Here, we reported the physiological significance of the newly identified general transcription termination factor of Archaea
Qi Zong Lao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 24(12), 5013-5023 (2010-08-25)
Voltage-gated calcium channels conduct Ca(2+) ions in response to membrane depolarization. The resulting transient increase in cytoplasmic free calcium concentration is a critical trigger for the initiation of such vital responses as muscle contraction and transcription. L-type Ca(v)1.2 calcium channels
Christoph Kalthoff et al.
The Journal of biological chemistry, 277(10), 8209-8216 (2002-01-05)
Epsin and AP180/CALM are important endocytic accessory proteins that are believed to be involved in the formation of clathrin coats. Both proteins associate with phosphorylated membrane inositol lipids through their epsin N-terminal homology domains and with other components of the
Maria Franzini et al.
Analytical biochemistry, 374(1), 1-6 (2007-11-21)
The clinical relevance of serum gamma-glutamyltransferase (GGT) activity, in areas other than hepatic function, has recently been increased by several epidemiological associations. Still, GGT remains a nonspecific test because of the influence of various pathophysiological factors. We devised a procedure
Germán L Rosano et al.
The Journal of biological chemistry, 286(34), 29671-29680 (2011-07-09)
HSP100 proteins are molecular chaperones involved in protein quality control. They assist in protein (un)folding, prevent aggregation, and are thought to participate in precursor translocation across membranes. Caseinolytic proteins ClpC and ClpD from plant chloroplasts belong to the HSP100 family.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service