Skip to Content
Merck
All Photos(3)

Key Documents

A8781

Sigma-Aldrich

β-Amylase from sweet potato

greener alternative

Synonym(s):

1,4-α-D-Glucan maltohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
41106514
NACRES:
NA.56

form

powder

mol wt

~200,000

packaging

vial of 15 mg

greener alternative product score

old score: 57
new score: 1
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Amylase from sweet potato is a tetrameric enzyme with each monomer comprising (α/β) core domain and a C-terminal loop region. It belongs to the glycoside hydrolase family GH14.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Application

β-Amylase from sweet potato has been used:
  • as a molecular weight marker in blue native polyacrylamide gel electrophoresis (PAGE)
  • as a standard for calibration curve preparation in size exclusion chromatography
  • as a model protein for small-angle X-ray analysis (SAXs)

Biochem/physiol Actions

β-Amylase acts on the polysaccharide α(1,4) glycosidic linkages and releases maltose present in the non-reducing ends. Due to its stability, β-Amylase from sweet potato is used in the immobilized enzyme preparation in combination with agarose beads.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integration of affinity precipitation with partitioning methods for bioseparation of chitin binding lectins.
Teotia, S. et al.
Food and Bioproducts Processing, 84, 37-43 (2006)
S Teotia et al.
Enzyme and microbial technology, 28(9-10), 792-795 (2001-06-09)
beta-amylases are used in production of maltose syrup. It is shown that sweet potato beta-amylase can be purified by affinity precipitation with alginate with 80% activity yield and 44 fold purification. SDS-PAGE of the purified protein showed a single band
Determination of hydrodynamic radius of proteins by size exclusion chromatography
LaVV, et al.
Bio-protocol, 7(8), 14-Jan-14-Jan (2017)
Purification of staphylococcal protein A by affinity precipitation using a reversibly soluble-insoluble polymer with human IgG as a ligand
Taniguchi, M. et al.
Journal of Fermentation and Bioengineering, 68, 32-36 (1989)
Khalilah G Reddie et al.
Molecular bioSystems, 4(6), 521-531 (2008-05-22)
Oxidation of the thiol functional group in cysteine (Cys-SH) to sulfenic (Cys-SOH), sulfinic (Cys-SO2H) and sulfonic acids (Cys-SO3H) is emerging as an important post-translational modification that can activate or deactivate the function of many proteins. Changes in thiol oxidation state

Protocols

Gel filtration chromatography is an established method for determining the size and molecular mass of proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service