Skip to Content
Merck

Skip To

MSCAL1

ProteoMass Peptide and Protein MALDI-MS Calibration Kit

Peptide and Protein MALDI MS Calibration Standard

Synonym(s):

MALDI-MS Calibration Kit for Proteins and Peptides

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ProteoMass

Quality Segment

quality

Peptide and Protein MALDI MS Calibration Standard

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Application

This kit features a convenient selection of pre-qualified standard peptides, proteins, matrices, and solvents for calibrating and testing matrix assisted laser desorption ionization (MALDI) mass spectrometers. Configured for analyzing complex mixtures of proteins and peptides (700 to 66,000 Da), MSCAL1 is ideal for evaluating unknown samples or samples containing a broad range of molecular weights. Applications include calibration, tuning, and sensitivity testing of MALDI instruments from all manufacturers.
“ProteoMass” Peptide and Protein MALDI-MS Calibration Kit has been used for calibration in the following experiments:
  • Isolation and characterization of cytotoxic cyclotides from Viola tricolor.(1)
  • Isolation and Characterization of novel cyclotides from Viola hederaceae.(2)
  • To study the domain architecture and biochemical characterization of vertebrate Mcm10.(3)

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSCAL3MSCAL2C8857
product line

ProteoMass

product line

ProteoMass

product line

ProteoMass

product line

ProteoMass

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

quality

Peptide and Protein MALDI MS Calibration Standard

quality

Protein MALDI MS Calibration Standard

quality

Peptide MALDI MS Calibration Standard

quality

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Kit Components Only

Product No.
Description

  • Sinapinic acid, Use tested for MALDI-TOF Mass Spec. 4 x 10

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 756.3997 Da 1 x 10
    SDS

  • ProteoMass P14R MALDI-MS Standard, vial of 10 nmol 1 x 10
    SDS

  • ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da 1 x 10
    SDS

  • ProteoMass Insulin chain B oxidized MALDI-MS Standard, vial of 10 nmol 1 x 10
    SDS

  • ProteoMass Insulin MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da 1 x 10
    SDS

  • ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation 1 x 10
    SDS

  • ProteoMass Apomyoglobin MALDI-MS Standard, vial of 10 nmol, (M+H+) 16,952.27 Da by calculation 1 x 10
    SDS

See All (12)

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



David C Ireland et al.
The Biochemical journal, 400(1), 1-12 (2006-07-29)
Cyclotides are a fascinating family of plant-derived peptides characterized by their head-to-tail cyclized backbone and knotted arrangement of three disulfide bonds. This conserved structural architecture, termed the CCK (cyclic cystine knot), is responsible for their exceptional resistance to thermal, chemical
Jun Yin et al.
British journal of pharmacology, 173(14), 2208-2223 (2016-06-25)
Chemical conjugation of therapeutic proteins with polyethylene glycol (PEG) is an established strategy to extend their biological half-life (t1/2 ) to a clinically useful range. We developed a novel uncharged and unstructured recombinant polypeptide composed of five amino acids (P
Cristina Pagliano et al.
Photosynthesis research, 108(1), 33-46 (2011-04-14)
We describe a one-step detergent solubilization protocol for isolating a highly active form of Photosystem II (PSII) from Pisum sativum L. Detailed characterization of the preparation showed that the complex was a monomer having no light harvesting proteins attached. This



Global Trade Item Number

SKUGTIN
MSCAL1-5X1KT04061833652633
MSCAL1-1KT04061835564262

Questions

  1. Good afternoon, I want to by this kit but I need more details about de name and mass of the peptides and proteins. Thank you very much.

    1 answer
    1. Components:
      All the standards supplied in the kit may also be purchased individually using the listed catalog numbers.
      Standard Peptides and Proteins:
      Standard peptides and proteins are supplied in 1.5 ml clear tubes, each containing 10 moles of standard.

      The details of the components of MSCAL1-1KT is attached.

      Helpful?

Reviews

No rating value

Active Filters