Skip to Content
Merck

Skip To

M3895

Mucin from bovine submaxillary glands

Type I-S

Synonym(s):

MUC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
282-357-4
NACRES:
NA.61
MDL number:
Form:
powder
Biological source:
bovine submaxillary glands
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine submaxillary glands

Quality Segment

type

Type I-S

form

powder

composition

Bound sialic acids, 9-24%

technique(s)

electrophoresis: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

cow ... BSM(286841)

General description

Neuraminidase substrate.
Mucin from bovine submaxillary glands is encoded by two bovine submaxillary BSM genes. It is present in the saliva and interacts with air and food. Mucin comprises three major domains namely N- and C terminal and the glycosylated proline, threonine and serine (PTS domain) central domain. The sialic acid contributes to 30% of the molecular weight of mucin. The central domain has many tandem repeats.

Application

Mucin from bovine submaxillary glands has been used:
  • in the quantification of mucin
  • as a component of artificial tear solution
  • as a substrate for recombinant sialidase Gardnerella vaginalis
  • to investigate a galactose-specific lectin from the red marine alga Ptilota filicina
  • to study the characterization of a unique mucin-like glycoprotein secreted by a human endometrial adenocarcinoma cell line (Ishikawa)

Biochem/physiol Actions

Albuminoid substance in mucus which gives it its ropy consistency
Bovine submaxillary mucin (BSM) is thermally stable up to 85°C. The terminal domain regions are key for the adsorption functionality of mucin.

Other Notes

Similar to M 4503, but produced by Sigma.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
499643L6385A8022
biological source

bovine submaxillary glands

biological source

-

biological source

bovine milk

biological source

bovine

technique(s)

electrophoresis: suitable

technique(s)

-

technique(s)

electrophoresis: suitable

technique(s)

ELISA: suitable, blocking: suitable, electrophoresis: suitable, western blot: suitable

form

powder

form

lyophilized

form

powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Gene Information

cow ... BSM(286841)

Gene Information

-

Gene Information

cow ... LALBA(281894)

Gene Information

bovine ... ALB(280717)

UniProt accession no.

P98091

UniProt accession no.

-

UniProt accession no.

P00711

UniProt accession no.

P02769


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Is this protein purified using His-tag or some other tag?

    1 answer
    1. The detailed protein purification process for this product is considered proprietary. This material is a native-source (non-recombinant) animal product with no affinity tag. The purification process is a modification of a CTAB-precipitation method of Tettamanti & Pigman (Arch. Biochem. Biophys. 124:41–50, 1968) and is then dialyzed against water (no buffer salts). The final pH is approximately 5–6. There are no buffer salts present in the final product and the material is not purified under native conditions.

      Pubmed link : https://pubmed.ncbi.nlm.nih.gov/5661614/

      Helpful?

  2. What is the recommended method to dissolve Mucin from bovine submaxillary glands, item M3895?

    1 answer
    1. Mucin from bovine submaxillary glands, item M3895, can be dissolved/reconstituted in H2O or dilute aqueous alkaline buffers.

      Helpful?

Reviews

No rating value

Active Filters