Skip to Content
Merck
All Photos(1)

Documents

M1942

Sigma-Aldrich

Molybdenum Blue spray reagent

liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

product name

Molybdenum Blue spray reagent,

description

1.3% molydbenum oxide in 4.2 M sulfuric acid
Molybdenum Blue reagent

Quality Level

form

liquid

technique(s)

thin layer chromatography (TLC): suitable

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

General description

Molybdenum Blue reagent is suitable for use in the detection of phospholipids and related compounds.

Application

For use in the detection of phospholipids and related compounds.
Molybdenum Blue reagent may be used for the determination of phosphatidylglycerol and the lecithin/sphingomyelin ratio in biological samples and phospholipids in Halobellus by using single-dimension thin-layer chromatography (TLC) method and high-performance thin-layer chromatography (HPTLC), respectively.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methods in Microbiology (1987)
G R Gotelli et al.
Clinical chemistry, 24(7), 1144-1146 (1978-07-01)
We describe a single-dimension thin-layer chromatographic method by which one can simultaneously determine the lecithin/sphingomyelin ratio and the proportion of phosphatidylglycerol in amniotic fluid. The phospholipids are conveniently detected by an improved molybdenum-blue spray reagent, which immediately produces blue spots
Tuan Manh Nguyen et al.
Journal of microbiology (Seoul, Korea), 55(8), 635-639 (2017-07-05)
There are marked differences between wet and freeze-dried cells with regard to the identification of polar lipid components. The determination of the polar lipid composition of freeze-dried cells is well established. However, several approaches to identifying polar lipid components in
Nocardioides panacisoli sp. nov., isolated from the soil of a ginseng field.
Cho CH
International Journal of Systematic and Evolutionary Microbiology, 60, 387-387 (2010)
Simona Lobasso et al.
Chemistry and physics of lipids, 186, 1-8 (2014-12-03)
Polar membrane lipids of an archaeal microorganism recently isolated from the natural salt lake Fuente de Piedra (Málaga, Spain) have been studied by means of TLC in combination with MALDI-TOF mass spectrometry. The major phospholipids are the ether lipids phosphatidylglycerophosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service