This product is supplied as a 2% (w/v) solution, essentially 2 grams per 100ml. Please see the link below to review the product datasheet:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/327/050/n7285pis.pdf
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352202
EC Number:
210-123-3
NACRES:
NA.56
Pricing and availability is not currently available.
Application
Ninhydrin reagents can be used in thin layer chromatography (TLC). Ninhydrin reagents have been used to develop and validate a simple, selective, and precise densitometric method for analysis of α-aminocephalosporins, both in bulk drugs and in formulations. Ninhydrin reagents have also been used to observe differences in the reference ranges of several plasma and urine amino acids between Singaporean and Caucasian populations.
Ninhydrin reagent has been used to confirm the presence of the amino acid moieties in various reactions.
Biochem/physiol Actions
Ninhydrin is a tricyclic 1,2,3-trione, which functions as an amino acid reagent. It is a vital organic building block, which exposes latent fingerprints on porous surfaces like paper, cardboard and raw wood. Ninhydrin is used to detect primary and secondary amines. It gives a strong blue color with primary amines and a less intense brown-red color with secondary amine.
Other Notes
Ninhydrin and hydrindantin in DMSO and lithium acetate buffer, pH 5.2
Disclaimer
Store under nitrogen in the refrigerator after use
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level 200 |
| form liquid | form solid | form solid | form powder |
| storage temp. 2-8°C | storage temp. 15-25°C | storage temp. - | storage temp. - |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
how to make 2 % ninhydrin solution
1 answer-
Helpful?
-
-
Please give preparation of 2% Ninhydrin solution
1 answer-
As listed on the Product Detail Page, Product N7285 is provided to the end-user as a 2% ninhydrin solution.
Helpful?
-


