Skip to Content
Merck
All Photos(3)

Documents

206865

Sigma-Aldrich

Primuline

Dye content 50 %

Synonym(s):

Direct Yellow 59, Primuline Yellow

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14N3NaO3S3
CAS Number:
Molecular Weight:
475.54
Colour Index Number:
49000
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder or crystals

Quality Level

composition

Dye content, 50%

mp

>300 °C

λmax

229 nm (2nd)
340-355 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Cc1ccc2nc(sc2c1S([O-])(=O)=O)-c3ccc4nc(sc4c3)-c5ccc(N)cc5

InChI

1S/C21H15N3O3S3.Na/c1-11-2-8-16-18(19(11)30(25,26)27)29-21(24-16)13-5-9-15-17(10-13)28-20(23-15)12-3-6-14(22)7-4-12;/h2-10H,22H2,1H3,(H,25,26,27);/q;+1/p-1

InChI key

RSRNHSYYBLEMOI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Primuline has been used as a reagent in the preparation of lipids for chloroplast lipid membrane. It has been used as a dye in preparative thin-layer chromatography (TLC).

Biochem/physiol Actions

Primuline, also known as direct yellow 59 or primuline yellow, is considered to be a more sensitive and non-destructive dye for lipids. It is widely used as a fluorescent dye in preparative thin-layer chromatography (TLC). Primuline is suitable for vital staining. It is used as a fluorochrome in plant tissues. Primuline is also used to stain pollen grains and lignified cell walls. In addition, it also helps in segregating intact and broken starch grains. At pH 8, primuline acts as a fluorochrome for mast cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Ndjomou et al.
Antiviral research, 96(2), 245-255 (2012-09-04)
The hepatitis C virus (HCV) multifunctional nonstructural protein 3 (NS3) is a protease that cleaves viral and host proteins and a helicase that separates DNA and RNA structures in reactions fueled by ATP hydrolysis. Li et al. (2012) recently synthesized
Sarah E Davis et al.
Infection and immunity, 86(8) (2018-06-06)
Candida albicans mutants for phosphatidylserine (PS) synthase (cho1ΔΔ) and PS decarboxylase (psd1ΔΔ psd2ΔΔ) are compromised for virulence in mouse models of systemic infection and oropharyngeal candidiasis (OPC). Both of these enzymes are necessary to synthesize phosphatidylethanolamine (PE) by the de
J L Labandeira Garcia et al.
Journal of anatomy, 137 (Pt 2), 247-261 (1983-09-01)
This paper describes a qualitative and quantitative investigation into the location of the motoneurons innervating the extra-ocular muscles of the rat. Injections of horseradish peroxidase, bisbenzimide, propidium iodide and DAPI-primuline were made either in one or simultaneously in two muscles.
V N Kazakov et al.
Neuroscience, 56(1), 157-164 (1993-09-01)
Retrograde axonal transport of fluorescent tracers (primuline, FluoroGold and Nuclear Yellow) from the spinal cord, frontal cortex, lateral hypothalamus to various neuronal groups of the midbrain periventricular gray substance (periaqueductal gray matter) and to the dorsolateral pontine tegmentum in the
Iris Meisen et al.
Analytical chemistry, 76(8), 2248-2255 (2004-04-15)
A combined strategy of preparative high-performance thin-layer chromatography overlay assay and mass spectrometry was established for the structural characterization of immunostained glycosphingolipids (GSLs) in silica gel extracts. Crude chloroform/methanol/water (30/60/8, v/v/v) extracts of immunostained TLC bands were analyzed by nanoelectrospray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service