Skip to Content
Merck
All Photos(1)

Key Documents

M1523

Sigma-Aldrich

Muscimol

≥98% (TLC), powder, GABAA receptor agonist.

Synonym(s):

3-Hydroxy-5-aminomethyl-isoxazole, 5-Aminomethyl-3-hydroxy-isoxazole, 5-Aminomethyl-3-isoxazolol, Agarine, Pantherine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
Beilstein:
1618960
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Muscimol, powder

form

powder

Quality Level

color

white to off-white

storage temp.

2-8°C

SMILES string

NCC1=CC(=O)NO1

InChI

1S/C4H6N2O2/c5-2-3-1-4(7)6-8-3/h1H,2,5H2,(H,6,7)

InChI key

ZJQHPWUVQPJPQT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Muscimol is a psychoactive isoxazole component, which is found in Amanita muscaria and related mushrooms.[1] It lowers cerebral glucose metabolism and increases the level of 5-hydroxytryptamine in brain.[2][3] Muscimol has hallucinogenic effects.[3] It is associated with distorted perceptions, nausea and vomiting.[4]

Application

Muscimol has been used:
  • to reduce conditioned fear-expression in lateral amygdala (LA)[5]
  • to eliminate cortical cell response in ferrets[6]
  • to inactivate lateral magnocellular nucleus of the nidopallium (LMAN) in zebra finches[7]

Biochem/physiol Actions

GABAA receptor agonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors and GABAC Receptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of the lateral amygdala in the retrieval and maintenance of fear-memories formed by repeated probabilistic reinforcement
Erlich JC, et al.
Frontiers in Behavioral Neuroscience, 6, 16-16 (2012)
Muscimol as an ionotropic GABA receptor agonist
Johnston GAR
Neurochemical Research, 39(10), 1942-1947 (2014)
Su-Hyuk Ko et al.
International journal of molecular sciences, 21(22) (2020-11-19)
Autophagy is a conserved pathway that plays a key role in cell homeostasis in normal settings, as well as abnormal and stress conditions. Autophagy dysfunction is found in various neurodegenerative diseases, although it remains unclear whether autophagy impairment is a
Julen Hernandez-Lallement et al.
Current biology : CB, 30(6), 949-961 (2020-03-07)
Empathy, the ability to share another individual's emotional state and/or experience, has been suggested to be a source of prosocial motivation by attributing negative value to actions that harm others. The neural underpinnings and evolution of such harm aversion remain
Handbook of Mushroom Poisoning: Diagnosis and Treatment (1994)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service