Skip to Content
Merck
All Photos(1)

Key Documents

A1304

Sigma-Aldrich

β-Amanitin

from Amanita phalloides, ≥85% (HPLC), powder, RNA polymerase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H53N9O15S
CAS Number:
Molecular Weight:
919.95
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

β-Amanitin from Amanita phalloides, ≥85% (HPLC)

Quality Level

Assay

≥85% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CCC(C)C1NC(=O)CNC(=O)C2Cc3c([nH]c4cc(O)ccc34)S(=O)CC(NC(=O)CNC1=O)C(=O)NC(CC(O)=O)C(=O)N5CC(O)CC5C(=O)NC(C(C)C(O)CO)C(=O)N2

InChI

1S/C39H53N9O15S/c1-4-16(2)31-36(60)41-11-28(53)42-25-15-64(63)38-21(20-6-5-18(50)7-22(20)45-38)9-23(33(57)40-12-29(54)46-31)43-37(61)32(17(3)27(52)14-49)47-35(59)26-8-19(51)13-48(26)39(62)24(10-30(55)56)44-34(25)58/h5-7,16-17,19,23-27,31-32,45,49-52H,4,8-15H2,1-3H3,(H,40,57)(H,41,60)(H,42,53)(H,43,61)(H,44,58)(H,46,54)(H,47,59)(H,55,56)

InChI key

IEQCUEXVAPAFMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Amanitin belongs to the family of amatoxins. Amanitins are made of bicyclic octapeptides. β-Amanitin is composed of a carboxyl group and is acidic in nature.

Application

β-Amanitin from Amanita phalloides has been used:
  • as a calibration standard for the quantification of β-Amanitin using liquid chromatography-high resolution-mass spectrometry/mass spectrometry (LC-HR-MS/MS) method.
  • to determine its concentration in urine samples by capillary zone electrophoresis (CZE).
  • in the analysis of β-amanitin in toxic mushrooms by liquid chromatography coupled to time-of-flight mass spectrometry.

Biochem/physiol Actions

Inhibits eukaryotic RNA polymerase II and III; does not inhibit RNA polymerase I or bacterial RNA polymerase; inhibits mammalian protein synthesis.
Toxic constituent of the mushroom, Amanita phalloides, inhibits eukaryotic RNA polymerase II and III, but not RNA polymerase I or bacterial RNA polymerase. Inhibits mammalian protein synthesis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Li et al.
Gene, 532(1), 63-71 (2013-09-21)
Amanita exitialis is a lethal mushroom that was first discovered in Guangdong Province, China. The high content of amanitin in its basidiocarps makes it lethal to humans. To comprehensively characterize the A. exitialis transcriptome and analyze the Amanita toxins as
Chiemi Nishizawa et al.
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 16(4), 441-445 (2004-01-27)
High-performance liquid chromatographic (HPLC) assay has been developed for the simultaneous determination of alpha-amanitin, beta-amanitin and phalloidin in serum. Three toxins were extracted by reflux in a water bath at 80 degrees C for one hour and purified by Sep-Pak
V A Robinson-Fuentes et al.
Journal of pharmaceutical and biomedical analysis, 47(4-5), 913-917 (2008-05-27)
Amanitins are toxins found in species of the mushroom genera Amanita, Lepiota and Galerina. Intoxication after ingestion of these mushrooms can be fatal with an estimated 20% of mortality rate. An early diagnosis is necessary in order to avoid invasive
H Faulstich et al.
Toxicon : official journal of the International Society on Toxinology, 26(5), 491-499 (1988-01-01)
A monoclonal antibody, with high affinity against the mushroom toxin alpha-amanitin, was prepared. Administration of the Fab fragment of the monoclonal antibody to mice caused a 50-fold increase in alpha-amanitin toxicity. Electron micrographs showed normal appearance of hepatocytes but typical
F Jehl et al.
Analytical biochemistry, 149(1), 35-42 (1985-08-15)
A high-performance liquid chromatographic assay of alpha-amanitin and beta-amanitin in human serum, urine, or stomach washings is described. Sample preparation involves a chemical step with deproteinization and organic solvent treatment, and a selective cleanup and concentration step on reversed-phase prepacked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service