Skip to Content
Merck
All Photos(1)

Key Documents

M0691

Sigma-Aldrich

Anti-Myosin VI antibody, Mouse monoclonal

~1 mg/mL, clone MUD-19, purified from hybridoma cell culture

Synonym(s):

Anti-DFNA22, Anti-DFNB37

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MUD-19, monoclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa by SDS-PAGE

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 2 μg/mL using A431 total cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO6(4646)
mouse ... Myo6(17920)
rat ... Myo6(315840)

General description

Monoclonal Anti-Myosin VI (mouse IgG1 isotype) is derived from the MUD-19 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with a synthetic peptide corresponding to amino acids of human myosin VI. Myosin VI is a relatively abundant widespread unconventional myosin composed of an N-terminal motor domain, a light chain binding neck region, a coil-coiled region and a highly conserved C-terminal domain.
Myosins are actin-dependent motors that have 15 classes based on their phylogenetics . Myosin VI is an unconventional actin motor that moves towards the minus ends of actin filaments, which is opposite to the movement of conventional myosins.

Specificity

Monoclonal Anti-Myosin VI antibody is specific for myosin VI in rats, humans and mice.

Application

Monoclonal Anti-Myosin VI antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • western blot assay
  • immunohistochemistry

Biochem/physiol Actions

Myosin VI is involved in the generation of cell shape change, cell motility, membrane remodeling and possibly in organelle and particle transport or tethering. It is also involved in membrane trafficking pathways in cultured mammalian cells where it is associated with the membrane ruffles and the trans-Golgi network. Myosin VI plays an essential role in the development of the mouse inner ear where it is expressed in the hair cell body, in the actin-rich cuticular plate at the base of the stereocilia and in the peri cuticular necklace region. Myosin VI gene mutations are involved in deafness and balance defects in the Snell′s Waltzer mouse. Myosin VI is present in the Golgi complex and may regulate vesicular transport across the membranes.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Wang et al.
Oncology reports, 36(3), 1285-1292 (2016-07-20)
Prostate cancer is the second most frequently diagnosed cancer among males around the world. Myosin VI (MYO6), as a motor protein, has been reported to be implicated in cancer-related cell migration and cellular functions. To investigate the role of MYO6 in
Unconventional Myosins in Inner-Ear Sensory Epithelia
Hasson T, et al.
The Journal of Cell Biology, 137(6), 1287-1287 (1997)
Rong Xu et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 20(2), 139-145 (2016-03-05)
Previously characterized as a backward motor, myosin VI (MYO6), which belongs to myosin family, moves toward the minus end of the actin track, a direction opposite to all other known myosin members. Recent researches have illuminated the role of MYO6
Francesca Demichelis et al.
Genes, chromosomes & cancer, 48(4), 366-380 (2009-01-22)
Emerging molecular and clinical data suggest that ETS fusion prostate cancer represents a distinct molecular subclass, driven most commonly by a hormonally regulated promoter and characterized by an aggressive natural history. The study of the genomic landscape of prostate cancer
MYO6 knockdown inhibits the growth and induces the apoptosis of prostate cancer cells by decreasing the phosphorylation of ERK1/2 and PRAS40
Wang D, et al.
Oncology Reports, 36(3), 1285-1292 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service