Skip to Content
Merck
All Photos(2)

Key Documents

M5187

Sigma-Aldrich

Anti-Myosin VI (KA-15) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DFNA22, Anti-DFNB37

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~150 kDa

species reactivity

rat, canine

technique(s)

indirect immunofluorescence: 1:75 using cultured rat NRK cells
microarray: suitable
western blot: 1:1,000 using a whole extract of cultured dog MDCK cells

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO6(4646)

General description

Myosin VI (MYO6) is localized to the Golgi complex and is expressed in the hair cells of the ear. It is a two-headed myosin with a short coiled-coil segment in its tail. The motor domain of MYO6 has two insertions. The gene encoding this protein is localized on chromosome 6q13.
Myosin VI is a relatively abundant widespread unconventional myosin composed of an N-terminal motor domain, a light chain binding neck region, a coiled-coiled region, and a highly conserved C-terminal domain. At the ′converter′ region, between the catalytic head and the neck region of Myosin VI, there is a characteristic linker about 50 amino acids long. Native Myosin VI is apparently a two-headed dimer of the heavy chains with each heavy chain bound to calmodulin light chain.

Immunogen

synthetic peptide corresponding to an epitope within the C-terminal of human Myosin VI, with N-terminal cysteine added, conjugated to KLH.

Application

Anti-Myosin VI (KA-15) antibody has been used:
  • in immunoblotting
  • in immunocytochemistry
  • in immunofluorescence
  • immunohistochemistry
  • proximity ligation assay

Biochem/physiol Actions

Myosin VI participates in the generation of cell shape change, cell motility, membrane remodeling, and possibly in organelle and particle transport or tethering. It is also involved in membrane trafficking pathways in cultured mammalian cells where it is associated with the membrane ruffles and the trans-Golgi network. The unusual direction of Myosin VI movement may suggest that it brings materials or membranes into the cell. Its activity in tissue cultured cells is thought to be regulated by phosphorylation. A mutation in Myosin VI was described recently in human autosomal dominant non syndromic hearing loss.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myosin VI is required for E-cadherin-mediated border cell migration
Geisbrecht E R, et al.
Nature Cell Biology, 4(8), 616-616 (2002)
Kali Burke et al.
eNeuro, 9(3) (2022-05-26)
Aging leads to degeneration of the peripheral and central auditory systems, hearing loss, and difficulty understanding sounds in noise. Aging is also associated with changes in susceptibility to or recovery from damaging noise exposures, although the effects of the interaction
Myosin VI: two distinct roles in endocytosis
Hasson T.
Journal of Cell Science, 116(17), 3453-3461 (2003)
Stepwise movements in vesicle transport of HER2 by motor proteins in living cells
Watanabe T M and Higuchi H
Biophysical Journal, 92(11), 4109-4120 (2007)
Shi-ming Yang et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 238(8), 1897-1908 (2009-07-08)
Smad4 is the central intracellular mediator of transforming growth factor-beta (TGF-beta) signaling, which plays crucial roles in tissue regeneration, cell differentiation, embryonic development, and regulation of the immune system. Conventional Smad4 gene knockout results in embryonic lethality, precluding its use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service