Skip to Content
Merck
All Photos(2)

Key Documents

D146005

Sigma-Aldrich

2,6-Dimethylaniline

99%

Synonym(s):

2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 20 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.560 (lit.)

bp

214 °C/739 mmHg (lit.)

mp

10-12 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)c1N

InChI

1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3

InChI key

UFFBMTHBGFGIHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,6-Dimethylaniline can react with:
  • 1-Naphthaldehyde to form an intermediate, bis(4-amino-3,5-dimethylphenyl)naphthylmethane (BADN) to further form bis(4-maleimido-3,5-dimethylphenyl)naphthylmethane (BMDN) by reacting with maleic anhydride, acetic anhydride and sodium acetate.
  • n-Butyllithium to form lithium 2,6-dimethylanilide, which can be utilized to form various RhII anilide complexes.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel bismaleimide with naphthalene side group. 1. From 1-naphthaldehyde and 2, 6-dimethylaniline
Wang CS, et al.
Polymer, 39(13), 2921-2927 (1998)
Kai Cheng et al.
Royal Society open science, 6(6), 190196-190196 (2019-07-18)
A novel aromatic diamine containing pyridyl side group, 4-pyridine-4,4-bis(3,5-dimethyl-5-aminophenyl)methane (PyDPM), was successfully synthesized via electrophilic substitution reaction. The polyimides (PIs) containing pyridine were obtained via the microwave-assisted one-step polycondensation of the PyDPM with pyromellitic dianhydride (PMDA), 3,3',4,4'-biphenyltetracarboxylic dianhydride (BPDA), 3,3',4,4'-diphenylether
Reactivity of Rhodium (II) Amido/Rhodium (I) Aminyl Complexes
Zhang N, et al.
Inorgorganica Chimica Acta, 39(13), 2921-2927 (2018)
Eva Tydén et al.
Toxicological sciences : an official journal of the Society of Toxicology, 81(2), 263-272 (2004-07-16)
Whole-body low-temperature radioluminography of (3)H-2,6-xylidine in rats indicates that the nonmetabolized substance, which is a volatile and fat-soluble compound, is distributed throughout the body and accumulates in adipose tissues, e.g., in the abdominal and subcutaneous regions. Whole-body autoradiography with freeze-dried
N W Puente et al.
Journal of analytical toxicology, 25(8), 711-715 (2002-01-05)
2,6-Dimethylaniline (2,6-xylidine; 2,6-DMA) is a nasal carcinogen in rats. Humans may be exposed to this compound via several routes: 2,6-DMA is found in cigarette smoke; it is a pharmacologically inactive metabolite of some drugs (e.g., the local anesthetic lidocaine) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service