Skip to Content
Merck
All Photos(1)

Key Documents

P9547

Sigma-Aldrich

Prilocaine hydrochloride

≥98% (TLC)

Synonym(s):

N-(2-Methylphenyl)-2-(propylamino)propanamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O · HCl
CAS Number:
Molecular Weight:
256.77
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

originator

AstraZeneca

SMILES string

Cl[H].CCCNC(C)C(=O)Nc1ccccc1C

InChI

1S/C13H20N2O.ClH/c1-4-9-14-11(3)13(16)15-12-8-6-5-7-10(12)2;/h5-8,11,14H,4,9H2,1-3H3,(H,15,16);1H

InChI key

BJPJNTKRKALCPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Used for local nerve block and for spinal anesthesia; pharmacological properties are similar to those of lidocaine.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Nakamura et al.
Canadian journal of anaesthesia = Journal canadien d'anesthesie, 46(5 Pt 1), 476-482 (1999-06-01)
To determine whether prilocaine, a local anesthetic, induces apoptosis in osteoblastic cells. After reaching subconfluence, human osteoblastic Saos-2 and MG63 cells and mouse osteoblastic MC3T3-E1 cells were exposed for 48 hr to varying concentrations of prilocaine up to 10 mM
Matthew A Petoe et al.
Journal of neurophysiology, 109(4), 1091-1096 (2012-12-12)
Temporary deafferentation of the upper limb, with ischemic or anesthetic nerve block, has rapid effects on sensorimotor cortex. Cutaneous anesthesia of the forearm has recently been found to improve sensory and motor function of the paretic hand in chronic stroke
Devendra Gupta et al.
Anesthesia and analgesia, 116(3), 568-571 (2013-02-13)
Topical capsaicin and eutectic mixture of local anesthetics (EMLA) have been found to be equally effective in minimizing the pain of venipuncture. After the injection of capsaicin, both tertiary amine local anesthetics and their quaternary ammonium derivatives can elicit a
Marco Bragagni et al.
International journal of pharmaceutics, 441(1-2), 468-475 (2012-11-21)
The use of injectable local anaesthetics for the treatment of severe postoperative pain is limited by the short duration of the painkilling effect. Pre-formulation studies were carried out for the development of an injectable microparticle formulation for controlled release of
Austin Larson et al.
Pediatric emergency care, 29(3), 377-379 (2013-03-07)
Topical anesthetics are commonly used in many health care settings and for many clinical conditions. However, there are a number of potential adverse effects associated with their use. Their widespread administration can convey a false sense of security and failure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service