Skip to Content
Merck
All Photos(1)

Key Documents

L5163

Sigma-Aldrich

Latrunculin A

from sea sponge, ≥85% (HPLC), waxy solid

Synonym(s):

LAT-A, [1R-[1R*, 4Z, 8E, 10Z, 12S*, 15R*, 17R*(R*)]]-4-(17-Hydroxy-5,12-dimethyl-3-oxo-2,16-dioxabicyclo[13.3.1]nonadeca-4,8,10-trien-17-yl)-2-thiazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO5S
CAS Number:
Molecular Weight:
421.55
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

sea sponge

Quality Level

Assay

≥85% (HPLC)

form

waxy solid

mol wt

421.6

solubility

DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

C[C@H]1CC[C@@H]2C[C@H](C[C@@](O)(O2)[C@@H]3CSC(=O)N3)OC(=O)C=C(C)CC\C=C\C=C1

InChI

1S/C22H31NO5S/c1-15-7-5-3-4-6-8-16(2)11-20(24)27-18-12-17(10-9-15)28-22(26,13-18)19-14-29-21(25)23-19/h3-5,7,11,15,17-19,26H,6,8-10,12-14H2,1-2H3,(H,23,25)/b4-3+,7-5-,16-11-/t15-,17-,18-,19+,22-/m1/s1

InChI key

DDVBPZROPPMBLW-IZGXTMSKSA-N

General description

Latrunculin A, a toxin extracted from the red sea sponge Latrunculia magnifica. It participates in vitro in the morphological alteration of the polymerization of pure actin. It forms a complex by binding with the nucleotide cleft of actin for actin filaments elongation.

Application

Latrunculin A has been used as medium supplementation for A549 cells to determine the internalization mechanism of CAV9 in A549 human lung carcinoma cells.

Biochem/physiol Actions

Latrunculin A inhibits actin polymerization by a different mechanism than cytochalasins. Latrunculin A disrupts microfilament-mediated processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maureen Möckel et al.
Journal of virology, 93(16) (2019-05-31)
Dynamin GTPases, best known for their role in membrane fission of endocytic vesicles, provide a target for viruses to be exploited during endocytic uptake. Recently, we found that entry of herpes simplex virus 1 (HSV-1) into skin cells depends on
M Coué et al.
FEBS letters, 213(2), 316-318 (1987-03-23)
Latrunculin A, a toxin purified from the red sea sponge Latrunculia magnifica, was found previously to induce striking reversible changes in the morphology of mammalian cells in culture and to disrupt the organization of their microfilaments. We now provide evidence
Keiichiro Kushiro et al.
Scientific reports, 7(1), 4244-4244 (2017-06-28)
During metastasis, cancer cells are exposed to various three-dimensional microstructures within the body, but the relationship between cancer migration and three-dimensional geometry remain largely unclear. Here, such geometric effects on cancerous cells were investigated by characterizing the motility of various
Peter J Wen et al.
Nature communications, 7, 12604-12604 (2016-09-01)
Vesicle fusion is executed via formation of an Ω-shaped structure (Ω-profile), followed by closure (kiss-and-run) or merging of the Ω-profile into the plasma membrane (full fusion). Although Ω-profile closure limits release but recycles vesicles economically, Ω-profile merging facilitates release but
Zhanghan Wu et al.
Nature communications, 9(1), 136-136 (2018-01-13)
Immune cells exhibit stimulation-dependent traveling waves in the cortex, much faster than typical cortical actin waves. These waves reflect rhythmic assembly of both actin machinery and peripheral membrane proteins such as F-BAR domain-containing proteins. Combining theory and experiments, we develop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service