Skip to Content
Merck
All Photos(1)

Key Documents

C6637

Sigma-Aldrich

Cytochalasin A from Drechslera dematioidea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H35NO5
CAS Number:
Molecular Weight:
477.59
Beilstein:
949521
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

−20°C

SMILES string

C[C@@H]1CCCC(=O)\C=C\C(=O)O[C@]23[C@@H](\C=C\C1)[C@H](O)C(=C)[C@@H](C)[C@H]2[C@H](Cc4ccccc4)NC3=O

InChI

1S/C29H35NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14-16,18-19,23-24,26-27,33H,3,7,9-10,13,17H2,1-2H3,(H,30,34)/b14-8+,16-15+/t18-,19-,23+,24+,26+,27-,29-/m1/s1

InChI key

ZMAODHOXRBLOQO-TZVKRXPSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Cytochalasin A reacts with sulfhydryls, and inhibits growth and glucose transport in Saccharomyces.
Cytochalasin A reacts with sulfhydryls, and inhibits growth and glucose transport in Saccharomyces. Cytochalasin A is an oxidizing analog of cytochalsin B.

Other Notes

Oxidized analog of cytochalasin B.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujiro Higuchi et al.
Biochemical and biophysical research communications, 340(3), 784-791 (2005-12-29)
Endocytosis is an important process for cellular activities. However, in filamentous fungi, the existence of endocytosis has been so far elusive. In this study, we used AoUapC-EGFP, the fusion protein of a putative uric acid-xanthine permease with enhanced green fluorescent
Tehila Hyman et al.
Molecular biology of the cell, 17(1), 427-437 (2005-10-28)
In epithelial cell lines, apical but not basolateral clathrin-mediated endocytosis has been shown to be affected by actin-disrupting drugs. Using electron and fluorescence microscopy, as well as biochemical assays, we show that the amount of actin dedicated to endocytosis is
Naimeh Taheri-Talesh et al.
Molecular biology of the cell, 19(4), 1439-1449 (2008-01-25)
Hyphal tip growth in fungi is important because of the economic and medical importance of fungi, and because it may be a useful model for polarized growth in other organisms. We have investigated the central questions of the roles of
Norio Takeshita et al.
Molecular biology of the cell, 16(4), 1961-1970 (2005-02-11)
One of the essential features of fungal morphogenesis is the polarized synthesis of cell wall components such as chitin. The actin cytoskeleton provides the structural basis for cell polarity in Aspergillus nidulans, as well as in most other eukaryotes. A
Kumiko Masai et al.
Bioscience, biotechnology, and biochemistry, 68(7), 1569-1573 (2004-07-28)
The distribution of the secreted protein ribonuclease T1 (RntA) fused with the enhanced green fluorescent protein (EGFP), RntA-EGFP, was visualized in hyphae of Aspergillus oryzae in the presence of a protein transport inhibitor, brefeldin A, cytochalasin A, or nocodazole. During

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service