Skip to Content
Merck
All Photos(1)

Key Documents

I8754

Sigma-Aldrich

L-allo-Isoleucine

≥99%

Synonym(s):

(2S,3R)-2-Amino-3-methylpentanoic acid, L-Alloisoleucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721791
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-allo-Isoleucine,

Assay

≥99%

Quality Level

form

powder

color

white to off-white

mp

285 °C

SMILES string

CC[C@@H](C)[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5+/m1/s1

InChI key

AGPKZVBTJJNPAG-UHNVWZDZSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Pseudomonas syringae pv. tomato DC3000 CmaL (PSPTO4723), a DUF1330 family member, is needed to produce L-allo-isoleucine, a precursor for the phytotoxin coronatine: This study demonstrates the essential role of L-allo-isoleucine in the synthesis of plant toxins, impacting plant pathology and biocontrol strategies (Worley et al., 2013).

Biochem/physiol Actions

L-allo-Isoleucine may be used in studies on maple syrup urine disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Wendel et al.
Pediatric research, 25(1), 11-14 (1989-01-01)
The nonprotein amino acid L-allo-isoleucine is formed endogenously in maple syrup urine disease patients from (R)-3-methyl-2-oxo-pentanoic acid. During strict metabolic balance, the plasma L-allo-isoleucine/L-isoleucine ratio correlates inversely with the residual activity of the branched-chain 2-oxoacid dehydrogenase in fibroblasts and thus
J E Baldwin et al.
The Biochemical journal, 301 ( Pt 2), 367-372 (1994-07-15)
Potential substrates for L-delta-(alpha-aminoadipoyl)-L-(cysteinyl)-D-valine (ACV) synthetase were initially identified using both the amino-acid-dependent ATP<-->pyrophosphate exchange reaction catalysed by the enzyme and the incorporation of 14C-radiolabelled cysteine and valine into potential peptide products. S-Carboxymethylcysteine was an effective substitute for alpha-aminoadipate and
P Schadewaldt et al.
Biochemical medicine and metabolic biology, 41(2), 105-116 (1989-04-01)
Possible functional differences in the catabolism of the four branched-chain L-amino acids in maple syrup urine disease were assessed using cultured human skin fibroblast stains. Transamination and oxidative decarboxylation were comparatively studied in 90-min incubations with 1 mmole/liter of 1-14C-labeled
Mahesha M Poojary et al.
Food chemistry, 234, 236-244 (2017-05-30)
In this study, enzyme-assisted extraction was performed to extract umami taste and total free amino acids (FAAs) from the six different mushrooms including shiitake (Lentinus edodes), oyster (Pleurotus ostreatus), tea tree (Agrocybe aegerita) and, white, brown and portobello champignons (Agaricus
Ana María Casas-Ferreira et al.
Talanta, 216, 120811-120811 (2020-05-28)
Herein we propose, for the first time, a rapid method based on flow injection analysis, electrospray ionization-tandem mass spectrometry (FIA-ESI-MS/MS) and multivariate calibration for the determination of l-leucine, l-isoleucine and L-allo-isoleucine in saliva. As far as we know, multivariate calibration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service