Skip to Content
Merck
All Photos(4)

Key Documents

I7403

Sigma-Aldrich

L-Isoleucine

98.5-101.0%, suitable for cell culture, BioXtra, non-animal source

Synonym(s):

(2S,3S)-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721792
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%

biological source

non-animal source

Quality Level

Agency

USP/NF
meets EP testing specifications
meets JP testing specifications
meets USP testing specifications

product line

BioXtra

Assay

98.5-101.0%

form

powder

quality

meets EP, JP, USP testing specifications

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

288 °C (dec.) (lit.)

solubility

1 M HCl: 50 mg/mL

application(s)

peptide synthesis
pharmaceutical (small molecule)

SMILES string

CC[C@H](C)[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Isoleucine has been used as a supplement of maintenance media consisting of DMEM/F-12 for the culture of primary hepatocytes. It has also been used as a supplement for serial culture of adult murine keratinocytes.

Biochem/physiol Actions

L-Isoleucine is an essential amino acid that is synthesized from threonine. It is a branched-chain hydrophobic amino acid. L-isoleucine is an isomer of L-leucine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibitory effect of aroma on the bitterness of branched-chain amino acid solutions.
Mukai J
Chemical & Pharmaceutical Bulletin, 55(11), 1581-1584 (2007)
Conversion of ammonia or urea into essential amino acids,L-leucine,L-valine, andL-isoleucine using artificial cells containing an immobilized multienzyme system and dextran-NAD+
Kang Fu Gu
Applied Biochemistry and Biotechnology, 26 (1990)
Biochemistry (5th Edition) null
Recapitulation of metabolic defects in a model of propionic acidemia using patient-derived primary hepatocytes.
Chapman KA
Molecular Genetics and Metabolism, 117(3), 355-362 (2016)
Serial Cultivation of Primary Adult Murine Keratinocytes
Richard P. Redvers and Pritinder Kaur
Methods in Molecular Biology (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service