Skip to Content
Merck
All Photos(3)

Key Documents

I7512

Sigma-Aldrich

Indole-3-butyric acid potassium salt

BioReagent, suitable for plant cell culture

Synonym(s):

4-(3-Indolyl)butanoic acid, IBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12KNO2
CAS Number:
Molecular Weight:
241.33
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

SMILES string

O=C(O[K])CCCC1=CNC2=C1C=CC=C2

InChI

1S/C12H13NO2.K.H/c14-12(15)7-3-4-9-8-13-11-6-2-1-5-10(9)11;;/h1-2,5-6,8,13H,3-4,7H2,(H,14,15);;

InChI key

RBRDCGZGWQSQJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Indole-3-butyric acid (IBA) is auxin-family plant hormone. IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cindy Kunze et al.
Nature communications, 8, 15858-15858 (2017-07-04)
The capacity of metal-containing porphyrinoids to mediate reductive dehalogenation is implemented in cobamide-containing reductive dehalogenases (RDases), which serve as terminal reductases in organohalide-respiring microbes. RDases allow for the exploitation of halogenated compounds as electron acceptors. Their reaction mechanism is under
Carolijn Smids et al.
Infection, 45(1), 41-49 (2016-06-19)
The purpose of this study was to evaluate the diagnostic value of From January 2006 to August 2013 patients with a clinical suspicion of spondylodiscitis, with an infection, or with fever of unknown origin were retrospectively included if Sixty-eight patients
Koen Vermeulen et al.
Theranostics, 9(2), 554-572 (2019-02-28)
Heat shock protein 90 is an ATP-dependent molecular chaperone important for folding, maturation and clearance of aberrantly expressed proteins and is abundantly expressed (1-2% of all proteins) in the cytosol of all normal cells. In some tumour cells, however, strong
Amy Midori James et al.
Plant physiology, 174(1), 154-171 (2017-03-30)
The accumulation of proanthocyanidins is regulated by a complex of transcription factors composed of R2R3 MYB, basic helix-loop-helix, and WD40 proteins that activate the promoters of biosynthetic genes. In poplar (genus
Jan M Schuller et al.
Science (New York, N.Y.), 363(6424), 257-260 (2018-12-24)
Photosynthetic complex I enables cyclic electron flow around photosystem I, a regulatory mechanism for photosynthetic energy conversion. We report a 3.3-angstrom-resolution cryo-electron microscopy structure of photosynthetic complex I from the cyanobacterium Thermosynechococcus elongatus. The model reveals structural adaptations that facilitate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service