Skip to Content
Merck
All Photos(1)

Key Documents

N1641

Sigma-Aldrich

1-Naphthylacetic acid

BioReagent, 1 mg/mL, suitable for plant cell culture

Synonym(s):

α-Naphthaleneacetic acid Free acid, 1-Naphthaleneacetic acid, NAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CO2H
CAS Number:
Molecular Weight:
186.21
Beilstein:
1308415
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

sterility

sterile-filtered

product line

BioReagent

form

solution

concentration

1 mg/mL

technique(s)

cell culture | plant: suitable

mp

129-131.5 °C (lit.)

application(s)

agriculture

storage temp.

2-8°C

SMILES string

OC(=O)Cc1cccc2ccccc12

InChI

1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)

InChI key

PRPINYUDVPFIRX-UHFFFAOYSA-N

Gene Information

Application

1-Naphthaleneacetic acid (NAA) is a synthetic phytohormone auxin that is added to cell culture media such as Murashige & Skoog media and Chu′s N6 media.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Qiang Chen et al.
Proteomics, 10(3), 369-379 (2009-12-24)
Although silk is used to produce textiles and serves as a valuable biomaterial in medicine, information on silk proteins of the cocoon is limited. Scanning electron microscopy was applied to morphologically characterise the sample and the solubility of cocoon in
Fatima Naim et al.
PloS one, 15(1), e0227994-e0227994 (2020-01-25)
Introducing a new trait into a crop through conventional breeding commonly takes decades, but recently developed genome sequence modification technology has the potential to accelerate this process. One of these new breeding technologies relies on an RNA-directed DNA nuclease (CRISPR/Cas9)
Michael A Q Martinez et al.
G3 (Bethesda, Md.), 10(1), 267-280 (2019-11-16)
As developmental biologists in the age of genome editing, we now have access to an ever-increasing array of tools to manipulate endogenous gene expression. The auxin-inducible degradation system allows for spatial and temporal control of protein degradation via a hormone-inducible
Jordi Chan et al.
Current biology : CB, 30(24), 4999-5006 (2020-10-10)
Several plant proteins are preferentially localized to one end of a cell, allowing a polarity to be assigned to the cell. These cell polarity proteins often exhibit coordinated patterns between neighboring cells, termed tissue cell polarity. Tissue cell polarity is
Rotem Matosevich et al.
Nature plants, 6(8), 1020-1030 (2020-08-05)
The root meristem can regenerate following removal of its stem-cell niche by recruitment of remnant cells from the stump. Regeneration is initiated by rapid accumulation of auxin near the injury site but the source of this auxin is unknown. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service