Skip to Content
Merck
All Photos(1)

Documents

I2521

Sigma-Aldrich

ITS+1 Liquid Media Supplement (100×)

liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Insulin-transferrin-sodium selenite, linoleic-BSA supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
NACRES:
NA.75

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

shipped in

ambient

storage temp.

2-8°C

General description

Serum-free culture environment avoids the variability and contaminants that can be introduced by serum. Serum-free formulations that substitute a purified form of the factors normally supplied by serum are suitable for many in vitro growth and differentiation studies. These factors include insulin, transferrin, selenium, pyruvate, and ethanolamine. The addition of other components varies greatly, depending on the cell type being studied and the basal medium employed. ITS is a mixture of insulin, human transferrin (partially iron-saturated), and sodium selenite. It is a general cell supplement designed for use in non-complex media (e.g., MEM, RPMI-1640) and complex media (e.g., Ham′s F-12, DME/F-12, MEM) with sodium pyruvate.

Application

ITS+1 Liquid Media Supplement (100×) has been used as a supplement in high-glucose Dulbecco′s Modified Eagle Medium (DMEM) for differentiation of mouse mesenchymal stem cells (mMSCs) to chondrocytes. It has also been used as a supplement in basal differentiation medium for differentiation of human-induced pluripotent stem cell (hiPSC) into osteoblasts under serum-free conditions.
Insulin-transferrin-sodium selenite, linoleic-BSA has been used for serum-free cell culture applications.ITS+1 Liquid Media Supplement (100×) has been used in culturing cell lines and for the preparation of immortalisation medium for the production of cell lines from ′Immortomouse′ embryos.

Components

Contains 1.0 mg/ml bovine insulin, 0.55 mg/ml human transferrin (substantially iron-free), 0.5 μg/ml sodium selenite, 50 mg/ml bovine serum albumin and 470 μg/ml linoleic acid at the 100x concentration.

Preparation Note

Prepared in Earle′s Balanced Salt Solution (EBSS) without phenol red.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heng Zhu et al.
Nature protocols, 5(3), 550-560 (2010-03-06)
Unlike humans, mouse bone marrow-derived mesenchymal stem cells (MSCs) cannot be easily harvested by adherence to plastic owing to the contamination of cultures by hematopoietic cells. The design of the protocol described here is based on the phenomenon that compact
Deniz M Özata et al.
Endocrine-related cancer, 18(6), 643-655 (2011-08-24)
Adrenocortical carcinoma (ACC) is an aggressive tumor showing frequent metastatic spread and poor survival. Although recent genome-wide studies of ACC have contributed to our understanding of the disease, major challenges remain for both diagnostic and prognostic assessments. The aim of
Jing Wen et al.
Scientific reports, 10(1), 2782-2782 (2020-02-19)
The developmental competence of IVM porcine oocytes is still low compared with that in their in vivo counterparts. Although many studies reported effects of glucose metabolism (GM) on oocyte nuclear maturation, few reported on cytoplasmic maturation. Previous studies could not
Phillip A Angart et al.
Applied microbiology and biotechnology, 101(14), 5645-5652 (2017-05-04)
Brain-derived neurotrophic factor (BDNF) is a member of the neurotrophin family critical for neuronal cell survival and differentiation, with therapeutic potential for the treatment of neurological disorders and spinal cord injuries. The production of recombinant, bioactive BDNF is not practical
Chang Guo et al.
Molecular nutrition & food research, e1801393-e1801393 (2019-06-07)
Alcoholic liver disease is a serious threat to human health. The development of drug candidates from complementary and alternative medicines is an attractive approach. Myricetin can be found in fruit, vegetables, and herbs. This study investigates the protective effect of

Articles

Importance and uses of linolenic acid in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures

Understand the role of selenium in cell culture media formulations. The Media Expert library explains how culture media ingredients support cell growth.

Understand the role of selenium in cell culture media formulations. The Media Expert library explains how culture media ingredients support cell growth.

Understand the role of selenium in cell culture media formulations. The Media Expert library explains how culture media ingredients support cell growth.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service