Skip to Content
Merck
All Photos(6)

Key Documents

HPA011880

Sigma-Aldrich

Anti-CUL4B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

CUL4B Antibody - Anti-CUL4B antibody produced in rabbit, Cul4B Antibody, Anti-CUL-4B antibody produced in rabbit, Anti-Cullin-4B antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GDGNDDEATTSKDGGFSSPSPSAAAAAQEVRSATDGNTSTTPPTSAKKRKLNSSSSSSSNSSNEREDFDSTSSSSSTPPLQPRDSASPSTSSFCLGVSVAASSHVPIQKKLRFEDTLEFVGFDAKMAEESSSSSSSSSPTAATS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL4B(8450)

Looking for similar products? Visit Product Comparison Guide

General description

Cullin 4B (CUL4B) gene is mapped to human chromosome Xq24. It codes for ubiquitin E3 ligase, which is a member of cullin-really interesting new gene (RING) ubiquitin ligase complex.

Specificity

Rabbit polyclonal anti-CUL4B antibody reacts with human Cullin-4B.

Immunogen

Cullin-4B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CUL4B antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Cullin 4B (CUL4B) plays an important development role in extra-embryonic tissue during mouse embryogenesis. It is implicated in regulation of various biological processes. CUL4B triggers tumor formation by inducing proliferation and preventing cell death of human osteosarcoma cells. Overexpression of the gene is associated with the development of non-small cell lung cancer (NSCLC). Therefore, knockdown ofCUL4B gene is considered to be a potential therapeutic method for treating NSCLC. Mutation in the gene leads to X-linked mental retardation (XLMR).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74400

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SIRT7 deacetylates DDB1 and suppresses the activity of the CRL4 E3 ligase complexes
Mo Y, et al.
FEBS Journal, 284(21), 3619-3636 (2017)
Knockdown of CUL4B Suppresses the proliferation and invasion in non-small cell lung cancer cells
Wang X and Chen Z
Oncology Research, 24(4), 271-277 (2016)
Hao Dou et al.
The Journal of biological chemistry, 294(48), 18504-18515 (2019-10-28)
Peroxisome proliferator-activated receptor γ (PPARγ) is the central regulator of adipogenesis, and its dysregulation is linked to obesity and metabolic diseases. Identification of the factors that regulate PPARγ expression and activity is therefore crucial for combating obesity. Aryl hydrocarbon receptor
Deletion of the CUL4B gene in a boy with mental retardation, minor facial anomalies, short stature, hypogonadism, and ataxia
Isidor B, et al.
American Journal of Medical Genetics. Part A, 152(1), 175-180 (2010)
Rescue of the genetically engineered Cul4b mutant mouse as a potential model for human X-linked mental retardation
Chen C, et al.
Human Molecular Genetics, 21(19), 4270-4285 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service