Skip to Content
Merck
All Photos(4)

Key Documents

MABE1084

Sigma-Aldrich

Anti-Topoisomerase I-DNA Covalent Complexes Antibody, clone 1.1A

clone 1.1A, from mouse

Synonym(s):

Topoisomerase I-DNA covalent complex, DNA topoisomerase 1-DNA covalent complex, Topo I-DNA covalent complex, TopoI cc, TopoIcc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1.1A, monoclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
dot blot: suitable
flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TOP1(7150)

General description

DNA topoisomerase 1 (EC 5.99.1.2; UniProt P11387; also known as DNA topoisomerase I, Topo I) is encoded by the TOP1 (also known as TOPI) gene (Gene ID 7150) in human. Topo I relaxes double strand DNA (dsDNA) torsional strain by nicking one strand of the dsDNA to allow rotation of the nicked 3′-hydroxy end around the unnicked strand, while the 5′-phosphoryl end of the nicked DNA forms a covalent phosphodiester bond with a tyrosine residue on Topo I. The nicked 3′-hydroxy end then attacks the DNA-enzyme phosphoester bond to religate the nicked DNA strand. One supercoil is removed during each round of Topo I-catalyzed nicking and resealing. Camptothecins are widely used for cancer treatment, these drugs intercalate into DNA at the topo I active site, inhibiting the religation step of the enzyme and shifting the equilibrium toward covalent topo I-DNA complexes rather than free topo I and DNA. Interaction of these drug-stabilized covalent topo I-DNA complexes with advancing replication forks or transcription complexes causse further DNA damage and eventual cell death.

Specificity

Clone 1.1A specifically recognizes topoisomerase I (Topo I) in DNA covalent complexes, but not non-DNA complexed, free Topo I by detecting topo I active site Tyr723 phosphorylation (Patel, A.G., et al, (2016). 44(6):2816-2826).

Immunogen

Epitope: Topo I pTyr723.
KLH-conjugated linear peptide corresponding to human Topo I target site sequence with phosphorylated Tyr723.

Application

Anti-Topoisomerase I-DNA Covalent Complexes Antibody, clone 1.1A is an antibody against Topoisomerase I-DNA Covalent Complexes for use in Immunocytochemistry, Flow Cytometry, Dot Blot, ELISA.
Flow Cytometry Analysis: 1.0 µg of this antibody from a representative lot detected topoisomerase I/DNA covalent complexes in Topotecan-treated HCT116 human colon cancer cells.

Immunocytochemistry Analysis: 10 µg/mL from a representative lot detected topoisomerase I/DNA covalent complexes in Topotecan-treated A549 human lung carcinoma cells (Courtesy of Dr. Scott H. Kaufmann, Mayo Clinic, Rochester, MN).

Flow Cytometry Analysis: 10 µg/mL from a representative lot detected topoisomerase I/DNA covalent complexes in Topotecan-treated HCT116 human colon cancer cells (Courtesy of Dr. Scott H. Kaufmann, Mayo Clinic, Rochester, MN).

Dot Blot Analysis: A representative lot detected the Topotecan-/TPT-stablized topoisomerase I/DNA covalent complexes, but not free topoisomerase I (non-DNA complexed) by slot blot using CsCl2 gradient-fractionated lysates from TPT-treated and untreated A549 cells (Patel, A.G., et al, (2016). 44(6):2816-2826).

Dot Blot Analysis: A representative lot detected a Topotecan (TPT) dose-dependent increase of TPT-stabilized topoisomerase I/DNA covalent complexes in lysates from TPT-treated A549 and HCT116 cells by slot blot (Patel, A.G., et al, (2016). 44(6):2816-2826).

Dot Blot Analysis: A representative lot detected stabilized topoisomerase I/DNA covalent complexes in lysates from A549 cells treated with Camptothecins (CPT, SN-38, TPT) or Indenoisoquinolines (NSC 314622, NSC 725776, NSC 743400), but not nucleoside analogues (cytarabine and gemcitabine) (Patel, A.G., et al, (2016). 44(6):2816-2826).

ELISA Analysis: A representative lot detected the immunogen peptide corresponding to topoisomerase I active site sequence with phosphorylated Tyr723, but not the peptide with non-phosphorylated Tyr273 by direct (non-sandwich) ELISA (Patel, A.G., et al, (2016). 44(6):2816-2826).

Immunocytochemistry Analysis: A representative lot detected a dose-dependent increase of non-nucleolar topoisomerase I/DNA covalent complex loci in Topotecan-/TPT-treated A549 cells, while the small number of loci in untreated cells were seen only in nucleoli (Patel, A.G., et al, (2016). 44(6):2816-2826).

Immunocytochemistry Analysis: A representative lot detected a temporally and spatially distinct nuclear loci formation of topoisomerase I/DNA covalent complexes from those of phospho-H2AX and Rad51 in Topotecan-/TPT-treated A549 cells (Patel, A.G., et al, (2016). 44(6):2816-2826).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Immunocytochemistry in Topotecan-treated A549 human lung carcinoma cells.

Immunocytochemistry Analysis: 10 µg/mL of this antibody detected topoisomerase I/DNA covalent complexes in Topotecan-treated A549 human lung carcinoma cells.

Target description

90.7 kDa calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Chieh Chiang et al.
Science advances, 3(4), e1602506-e1602506 (2017-05-17)
Breakage of one strand of DNA is the most common form of DNA damage. Most damaged DNA termini require end-processing in preparation for ligation. The importance of this step is highlighted by the association of defects in the 3'-end processing
Naoto Shimizu et al.
The Journal of biological chemistry, 299(8), 104988-104988 (2023-07-02)
Topoisomerases are enzymes that relax DNA supercoiling during replication and transcription. Camptothecin, a topoisomerase 1 (TOP1) inhibitor, and its analogs trap TOP1 at the 3'-end of DNA as a DNA-bound intermediate, resulting in DNA damage that can kill cells. Drugs
Ignacio Torrecilla et al.
Nucleic acids research, 52(2), 525-547 (2023-12-12)
DNA-protein crosslinks (DPCs) are toxic DNA lesions wherein a protein is covalently attached to DNA. If not rapidly repaired, DPCs create obstacles that disturb DNA replication, transcription and DNA damage repair, ultimately leading to genome instability. The persistence of DPCs
Angela M Mabb et al.
PloS one, 11(5), e0156439-e0156439 (2016-05-28)
Topoisomerase 1 (TOP1) inhibitors, including camptothecin and topotecan, covalently trap TOP1 on DNA, creating cleavage complexes (cc's) that must be resolved before gene transcription and DNA replication can proceed. We previously found that topotecan reduces the expression of long (>100
Osama Hidmi et al.
iScience, 27(3), 109082-109082 (2024-02-20)
DNA double-stranded breaks (DSBs) pose a significant threat to genomic integrity, and their generation during essential cellular processes like transcription remains poorly understood. In this study, we employ several techniques to map DSBs, R-loops, and topoisomerase 1 cleavage complex (TOP1cc)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service