Skip to Content
Merck

Skip To

D5207

Doxycycline hyclate

powder, suitable for cell culture, BioReagent

Synonym(s):

Doxycycline hyclate, α-6-Deoxy-5-hydroxytetracycline, Doxycycline hydrochloride hemiethanolate hemihydrate, Vibramycin Hyclate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
UNSPSC Code:
12352207
NACRES:
NA.22
MDL number:
Beilstein/REAXYS Number:
5702728
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Doxycycline hyclate, Suitable for Cell Culture

Quality Segment

form

powder

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].[H]O[H].CCO.C[C@@H]1C2[C@H](O)C3[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C2C(=O)c4c(O)cccc14.C[C@@H]5C6[C@H](O)C7[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]7(O)C(O)=C6C(=O)c8c(O)cccc58

InChI

1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10?,14?,15-,17-,22-;;;;/m00..../s1

InChI key

HALQELOKLVRWRI-ZVACAFRPSA-N

General description

General description
Chemical structure: tetracycline

Doxycycline hyclate belongs to the second generation of tetracycline antibiotics family. Doxycycline hyclate shows minimal side effects upon usage. It is effective for the treatment of recurrent aphthous stomatitis (RAS) and promotes speedy recovery.

Application

Doxycycline hyclate has been used for the induction of glioma cells and human embryonic kidney 293T cells (HEK293T).

Doxycycline hyclate is a synthetic oxytetracycline derivative. It has been used to eliminate Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs and to eliminate Ixodes scapularis ticks. It is a broad spectrum inhibitor used to inhibit matrix metalloproteinases (MMP), such as type 1 collagenase in studies on wound healing and tissue remodeling.

Biochem/physiol Actions

Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi. It is a broad spectrum inhibitor of matrix metalloproteinases in vivo.
Mode of Action: Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi.

Activity Spectrum: Effective against a broad spectrum inhibitor of matrix metalloproteinases in vivo.

Packaging

1, 5, 10 g in glass bottle

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR114533429D3000000
description

Suitable for Cell Culture

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

VETRANAL®, analytical standard

description

European Pharmacopoeia (EP) Reference Standard

Quality Level

200

Quality Level

300

Quality Level

100

Quality Level

-

form

powder

form

-

form

-

form

-

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

-

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Doxycycline hyclate


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. If I have made 50mg/ml solution in water using this doxycycline powder, then where to store that solution?

    1 answer
    1. Aqueous solutions of Doxycycline are light sensitive and should be used within 48 hours. The long term stability of aqueous solutions at -20°C has not been evaluated. However, some published articles indicate that doxycycline remained stable for 7 days in acidified water for 14 days. Another source states that solutions of doxycycline hyclate at a concentration of 10 mg/mL in sterile water, when frozen immediately after reconstitution, are stable for 8 weeks when stored at −20°C. Please see the link below to review one such article. Additional information may be available, consulting the relevant literature is advisable:
      https://pmc.ncbi.nlm.nih.gov/articles/PMC4943619/
      Please see the following link to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/402/297/d5207dat.pdf

      Helpful?

  2. Question related to Stephane F's and the answer. I resuspended in DMSO but the solution that I have is slightly yellow is that okay? Also how long does the powder last at 4C?

    1 answer
    1. This product does not have a color specification for the solution upon solubilizing. It is soluble in water (50 mg/ml) and in DMSO (10 mg/ml), yielding a clear solution. Mild warming may be required to fully dissolve the material. If the solution is clear and slightly yellowish, it should be acceptable for use.
      The recommended storage temperature for this product is 2–8 °C. If unopened and stored at 4–8 °C, it is guaranteed for 1 year from the date of shipment. Additional information is available in the product datasheet: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/402/297/d5207dat.pdf

      Helpful?

  3. What temperature should a solution be stored?

    1 answer
    1. As stated in the 'PROPERTIES' section, the recommended storage temperature for this product is 2–8 °C. Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/402/297/d5207dat.pdf

      Helpful?

  4. Is this product suitable for in-vivo use (added to drinking water)? Also what is the difference between this product (D5207) and D9891? thank you

    1 answer
    1. This product is intended for research use only. It is not suitable for human or veterinary use. The material has not been tested in animal models for ingestion. The end user would need to determine suitability. The major difference between product D5207 and D9891 is the testing performed. Product D5207 is tested suitable for cell culture applications. Please see the product specification sheet below regarding testing:
      https://www.sigmaaldrich.com/specification-sheets/330/944/D5207-10G.pdf
      https://www.sigmaaldrich.com/specification-sheets/323/301/D9891-25G.pdf

      Helpful?

  5. Hello. I have a question regarding the ref D5207 Doxycycline hyclate. I would like to know how I can resuspend the powder (DMSO, H2O, etc...). And at which concentration. Is that 10 mg/ml ok ? Thank you.

    1 answer
    1. This product is soluble in water (50 mg/mL), and in DMSO (10 mg/mL) yielding a clear solution. Mild warming may be required to fully dissolve the material.

      Please see the product data sheet for more information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/402/297/d5207dat.pdf

      Helpful?

Reviews

No rating value

Active Filters