Skip to Content
Merck
All Photos(5)

Key Documents

HPA004471

Sigma-Aldrich

Anti-KIT antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD117 antigen, Anti-Mast/stem cell growth factor receptor precursor, Anti-Proto-oncogene tyrosine-protein kinase Kit, Anti-SCFR, Anti-c-kit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

VGDEIRLLCTDPGFVKWTFEILDETNENKQNEWITEKAEATNTGKYTCTNKHGLSNSIYVFVRDPAKLFLVDRSLYGKEDNDTLVRCPLTDPEVTNYSLKGCQGKPLPKDLRFIPDPKAGIMIKSVKRAYHRLCLHCSVDQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIT(3815)

General description

The KIT/c-KIT proto-oncogene encodes a type III tyrosine kinase receptor. The KIT gene is located on the human chromosome at 4q12.

Immunogen

Mast/stem cell growth factor receptor precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-KIT antibody produced in rabbit has been used in:
  • immunohistochemistry
  • fluorescence microscopy
  • immunofluorescence

Biochem/physiol Actions

KIT tyrosine kinase receptor is involved in the survival and maintenance of hematopoietic cells, germ cells, and melanocytes. In association with protein tyrosine phosphatase receptor omicron (PTP-RO), KIT plays an important role in the megakaryocytopoiesis. Mutations in the KIT gene are associated with acute myeloid leukemia (AML).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86859

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Joanna Baxter et al.
Human molecular genetics, 11(12), 1391-1397 (2002-05-25)
Chronic myeloid leukaemia (CML) is characterized by the presence of the BCR-ABL fusion gene, usually in association with the t(9;22)(q34;q11) translocation. We report here the identification and cloning of a rare variant translocation, t(4;22)(q12;q11), in two patients with a CML-like
M Kossai et al.
Lung cancer (Amsterdam, Netherlands), 89(3), 306-310 (2015-07-15)
Thymic epithelial neoplasms (TENs) represent a rare entity with poor prognosis and limited systemic treatment options. The aim of this study was to assess the clinical benefit, the efficacy and toxicities of agents for patients with TEN enrolled in Phase
Innocenzo Sammarco et al.
International journal of colorectal disease, 19(6), 545-553 (2004-05-11)
The proto-oncogene c-KIT encodes a tyrosine kinase receptor essential during embryonic development and postnatal life. Although deregulated expression of c-KIT has been reported, its role in colorectal carcinoma remains controversial: some authors have described a correlation between c-KIT expression and
Xiao-ning Gao et al.
Leukemia research, 35(9), 1226-1232 (2011-07-05)
Mutations and/or overexpression of c-Kit proto-oncogene frequently occur in subsets of acute myeloid leukemia (AML) and contribute to abnormal cell proliferation and poor outcomes. We showed that c-Kit expression was subject to post-transcriptional regulation by microRNA (miRNA)-193b. Notably, miR-193b was
Jason Lowery et al.
The Journal of biological chemistry, 286(42), 36898-36906 (2011-08-11)
ADP-ribosylation factors (ARFs) and their activating guanine nucleotide exchange factors (GEFs) play key roles in membrane traffic and signaling. All ARF GEFs share a ∼200-residue Sec7 domain (Sec7d) that alone catalyzes the GDP to GTP exchange that activates ARF. We

Articles

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Mesenchymal stem cell markers and antibodies suitable for investigating targets in fibroblasts, chondrocytes, adipocytes, osteoblasts, and muscle cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service