Skip to Content
Merck

Skip To

MABE167

Anti-SNAI1 Antibody, clone 10H4.1

clone 10H4.1, from mouse

Synonym(s):

Zinc finger protein SNAI1, Protein snail homolog 1, Protein sna

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
10H4.1, monoclonal
Application:
IHC, WB
Citations:
6
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10H4.1, monoclonal

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry: suitable, western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SNAI1(6615)

General description

SNAI1, also known as Snail homolog 1, is a transcription factor that down regulates the expression of ectodermal genes within the mesoderm and is thought to be essential for mesoderm formation in the developing embryo. SNAI1 is expressed in the kidney and in mesenchymal and epithelial cell lines. Mutations in SNAI1 have been associated with tumors.
~29 kDa observed

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of Human SNAIL1.

Application

Anti-SNAI1 Antibody, clone 10H4.1 is a highly specific mouse monoclonal antibody, that targets Zinc Finger Protein & has been tested in western blotting & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected SNAIL1 in 10 µg of human and mouse kidney tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected SNAIL1 in human kidney section and human tonsil section tissues.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in rat kidney tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected SNAIL1 in 10 µg of rat kidney tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABD38SAB1404386SAB1401367
clone

10H4.1, monoclonal

clone

polyclonal

clone

2G11, monoclonal

clone

1A5, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

species reactivity

human, rat, mouse

species reactivity

mouse, rat

species reactivity

human

species reactivity

human

Gene Information

human ... SNAI1(6615)

Gene Information

human ... SNAI1(6615)

Gene Information

human ... SNAI1(6615)

Gene Information

human ... SNAI1(6615)

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters