Skip to Content
Merck
All Photos(5)

Documents

HPA001426

Sigma-Aldrich

Anti-LGMN antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Asparaginyl endopeptidase antibody produced in rabbit, Anti-Legumain precursor antibody produced in rabbit, Anti-Protease, cysteine 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

QGMKRKASSPVPLPPVTHLDLTPSPDVPLTIMKRKLMNTNDLEESRQLTEEIQRHLDARHLIEKSVRKIVSLLAASEAEVEQLLSERAPLTGHSCYPEALLHFRTHCFNWHSPTYEYALRHLYVLVNLCEKPYPLHRIKLSMDHVCLGHY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LGMN(5641)

General description

LGMN (legumain) is a cysteine lysosomal endoprotease that cleaves substrates at specific aspartate or asparagine residues. It is predominantly expressed in placenta and kidney, and is also found in brain, heart, liver, spleen and testis. It has a caspase-like fold, with a central six-stranded β-sheet (β1–β6), flanked by five major α-helices (α1–α5). The active form is a monomer. The enzyme is activated autocatalytically by acidic pH. The gene is localized to chromosome 14q32.1.

Immunogen

Legumain precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

LGMN (Legumain) gene encodes a cysteine protease that specifically catalyzes the hydrolysis of asparaginyl bonds. It is involved in the processing of proteins (bacterial and endogenous) for MHC class II presentation in the lysosomal/endosomal systems. Overexpression of legumain has been observed in several solid tumors. It serves as a prognostic marker and a potential therapeutic target is certain cancers, such as gastric carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70368

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Tanaka et al.
Cytogenetics and cell genetics, 74(1-2), 120-123 (1996-01-01)
We have isolated a novel human cDNA encoding a protein of 433 amino acids which shows 40% sequence identity to a hemoglobinase of Schistosoma japonicum, one of the cysteine proteases in the pathway by which trematodes degrade host-cell globin. It
Pengtao Guo et al.
PloS one, 8(9), e73090-e73090 (2013-09-12)
Legumain, a novel asparaginyl endopeptidase, has been observed to be highly expressed in several types of tumors, which may play a vital role in carcinogenesis. However, there is no study investigating the relationship among Legumain expression, clinicopathologic, biological variables and
Lixia Zhao et al.
Cell research, 24(3), 344-358 (2014-01-11)
Asparaginyl endopeptidase (AEP) is an endo/lysosomal cysteine endopeptidase with a preference for an asparagine residue at the P1 site and plays an important role in the maturation of toll-like receptors 3/7/9. AEP is known to undergo autoproteolytic maturation at acidic
Elfriede Dall et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(27), 10940-10945 (2013-06-19)
The cysteine protease legumain plays important functions in immunity and cancer at different cellular locations, some of which appeared conflicting with its proteolytic activity and stability. Here, we report crystal structures of legumain in the zymogenic and fully activated form
Tong Wu et al.
Molecular medicine reports, 13(3), 2377-2384 (2016-02-06)
The present study aimed to assess the expression of legumain in uveal melanoma (UM) cell lines and primary UM specimens, and to determine the possible association between legumain expression and clinical as well as pathological characteristics to reveal its impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service