Skip to Content
Merck
All Photos(1)

Key Documents

D5921

Sigma-Aldrich

DMEM - low glucose

With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: no
sodium pyruvate: no
glucose: low
HEPES: no
L-glutamine: no
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Dulbecco′s Modified Eagle′s medium (DME) is a modification of Basal Medium Eagle (BME), which has a four-fold higher concentration of amino acids and vitamins along with additional supplementary components. The first original DME formula was reported to culture embryonic mouse cells comprising 1,000 mg/L of glucose. An alteration with 4,500 mg/L glucose is optimal in cultivating certain cell types. DME has been shown to improve the stability of the medium.

Application

Dulbecco′s Modified Eagle′s medium-low glucose has been used to culture:
  • living colon cancer SW620 cells
  • human breast carcinoma cell lines MCF-7(13), KPL and MDA-MB-231
  • mammal and canine derived adipose-derived mesenchymal stem cells

Reconstitution

Supplement with 0.584 g/L L-glutamine.

Other Notes

Investigators who work with stem cells or cell clones frequently prefer media without phenol red.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Ohe et al.
The Journal of steroid biochemistry and molecular biology, 182, 21-26 (2018-04-22)
The high-mobility group A protein 1a (HMGA1a) protein is known as an oncogene whose expression level in cancer tissue correlates with the malignant potential, and known as a component of senescence-related structures connecting it to tumor suppressor networks in fibroblasts.
C Mignon et al.
Scientific reports, 7(1), 2797-2797 (2017-06-07)
Photobiomodulation-based (LLLT) therapies show tantalizing promise for treatment of skin diseases. Confidence in this approach is blighted however by lamentable inconsistency in published experimental designs, and so complicates interpretation. Here we interrogate the appropriateness of a range of previously-reported treatment
Tao Dong et al.
Molecular human reproduction, 26(6), 441-451 (2020-04-26)
Concentrations of 2-methoxyestradiol (2ME2), a principal metabolite of estradiol, are significantly lower in women with severe preeclampsia. Nitric oxide (NO) released by endothelial nitric oxide synthase (eNOS) plays an important role in regulating cardiovascular homeostasis. Importantly, high-density lipoprotein (HDL) stimulates
Giuseppe Antonacci et al.
Scientific reports, 6, 37217-37217 (2016-11-16)
Cellular biomechanics play a pivotal role in the pathophysiology of several diseases. Unfortunately, current methods to measure biomechanical properties are invasive and mostly limited to the surface of a cell. As a result, the mechanical behaviour of subcellular structures and
Christina Schmid et al.
Journal of pharmacological and toxicological methods, 106, 106915-106915 (2020-09-02)
Human induced pluripotent stem cell (hiPSC)-derived cardiomyocytes are being evaluated for their use in pharmacological and toxicological testing, particularly for electrophysiological side effects. However, little is known about the composition of the commercially available iCell cardiomyocyte (Fuijifilm Cellular Dynamics) cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service