Skip to Content
Merck

Skip To

D8537

Dulbecco’s Phosphate Buffered Saline

liquid, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture

1 of 6 reviewers received a sample product or took part in a promotion

Synonym(s):

DPBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
eCl@ss:
32129211
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: no (Dextro)
sodium pyruvate: no

shipped in

ambient

storage temp.

room temp

General description

Dulbecco’s Phosphate Buffered Saline (DPBS) is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. DPBS buffer is used to irrigate, wash, and dilute mammalian cells while maintaining osmotic balance. It provides water and inorganic ions essential for normal cell metabolism. DPBS with calcium and magnesium facilitates cell binding and clumping. Dulbecco′s PBS without these ions can be used to wash and rinse suspended cells. Phosphate buffering maintains the pH in the physiological range.

Application

Dulbecco′s Phosphate Buffered Saline has been used:

  • for the dilution of fresh red blood cells to study the trapping of red blood cell aggregate.
  • to wash the wells of a microtiter plate.
  • to wash the cells before resuspending them in lysis buffer for DNA isolation.
  • as a component of embryo holding medium for vitrification of embryos.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
59321C59331C59300C
description

Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture

description

liquid, sterile-filtered, DPBS Modified, without calcium, without magnesium, suitable for cell culture

description

liquid, sterile-filtered, DPBS Modified 10X, without calcium, without magnesium, suitable for cell culture

description

liquid, sterile-filtered, with calcium, with magnesium, suitable for cell culture

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

Quality Level

300

Quality Level

400

Quality Level

400

Quality Level

500

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

room temp

storage temp.

-

storage temp.

-

storage temp.

room temp

impurities

endotoxin, tested

impurities

-

impurities

-

impurities

-

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 11 Questions  
  1. Eu gostaria de saber a osmolaridade da solução de DMPBS.

    1 answer
    1. This product has an osmolarity range of 275–304 mOsm/kg.

      Helpful?

  2. Quería saber si éste producto se puede considerar RNAsa free para realizar durante la hibridación in situ

    1 answer
    1. This product is a sterile-filtered M Quality Level 300 product. It is not molecular biology grade and does not have specifications such as RNase and DNase free. Therefore, this product cannot be considered RNase free for use in in situ hybridization.

      Helpful?

  3. Bonjour, pourriez-vous m'indiquer le numéro de CAS de la solution de phosphate saline dulbecco sous forme liquide

    1 answer
    1. This item is a multi-component product. A CAS number cannot be applied to this type of product. The Safety Data Sheet document includes the individual CAS numbers of those components that may be potentially hazardous.

      Helpful?

  4. Hi, is this product GMP-grade or are GMP-garde material is used for the production of this?

    1 answer
    1. This product is not GMP-grade. Unfortunately. a GMP-grade version of this product is not available.

      Helpful?

  5. What is the concentration of the product in Molarity?

    1 answer
    1. This is a multi-component formulation. A 'molarity' is not reported for this type of product. See below for the list of components:
      Product D8537 contains:
      Potassium chloride: 0.2 g/l
      Potassium phosphate monobasic: 0.2 g/l
      Sodium chloride: 8 g/l
      Sodium Phosphate dibasic: 1.15 g/l

      Please see the link below for additional information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/124/464/d8537for.pdf

      Helpful?

  6. What is the bottle thread type on the 1L bottle (D8537-1L)

    1 answer
    1. The product is packaged in a square PET bottle with an HDPE cap. The cap specifications are as follows:
      Thread Type 38-430
      Thread Diameter 37.3 mm

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. What is the formulation for Product D8537, Dulbecco's Phosphate?

    1 answer
    1. The formulation for 1 liter is 8 g Sodium Chloride, 0.2 g Potassium Phosphate, monobasic, 1.15 g Sodium Phosphate, dibasic, and 0.2 g Potassium Chloride.

      Helpful?

  9. What is the pH of a solution of Product D8537, Dulbecco's Phosphate?

    1 answer
    1. The pH will be in the range of 7.1-7.5, according to the specification sheet.

      Helpful?

  10. Is Product D8537, Dulbecco's Phosphate, a 1X or 10 X concentration?

    1 answer
    1. This is a 1X concentration. The 10X concentration is listed under product D1408.

      Helpful?

1–10 of 11 Questions  

Reviews

Active Filters

  1. Atlanta, GA
    • Reviews 3
    • Votes 104
    5 out of 5 stars.

    All-purpose DPBS

    It's DPBS with calcium-magnesium modification. Worked pretty well for me for various applications.

    Helpful?

  2. 5 Ratings-only reviews