Skip to Content
Merck
All Photos(8)

Key Documents

HPA000264

Sigma-Aldrich

Anti-ETV6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ETS translocation variant 6 antibody produced in rabbit, Anti-ETS-related protein Tel1 antibody produced in rabbit, Anti-Tel antibody produced in rabbit, Anti-Transcription factor ETV6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NGKALLLLTKEDFRYRSPHSGDVLYELLQHILKQRKPRILFSPFFHPGNSIHTQPEVILHQNHEEDNCVQRTPRPSVDNVHHNPPTIELLHRSRSPITTNHRPSPDPEQRPLRSPLDNMIR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ETV6(2120)

General description

Ets variant gene 6 (ETV6) is mapped to human chromosome 12p13. ETV6 protein consists of two domains namely, the HLH (helix-loop-helix) domain and the ETS (E26 transformation-specific) domain encoded by exons 3 and 4 and exons 6 through 8, respectively. It also contains an internal domain encoded by exon 5.

Immunogen

Transcription factor ETV6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ETV6-NTRK3 (neurotrophic tyrosine kinase receptor type 3) gene fusions might function as a diagnostic marker for congenital (or infantile) fibrosarcoma (CFS). They also play a vital role in building a histogenetic relationship between CFS and congenital mesoblastic nephroma (CMN). ETV6 gene selectively regulates postembryonic hematopoietic stem cells (HSCs) survival. The encoded protein acts as an active transcriptional repressor, by means of its helix-loop-helix (HLH) and internal domains. ETV6 might be involved in modification of a transcription factor function, activation of kinase activity of partner gene and can cause leukemogenesis by its multiple rearrangement partner genes. ETV6 might contribute to various pathogenic mechanisms, leading to cell transformation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76166

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah W Sevilla et al.
Human pathology, 40(4), 588-593 (2008-11-11)
Cytogenetic abnormalities of chromosome 12p involving the TEL/ETV6 gene are observed in a variety of hematopoietic neoplasms including acute leukemias, myelodysplastic syndromes, and myeloproliferative disorders. Karyotypic aberrations, including rearrangements, deletions, and amplifications of chromosome 12p, have been documented in B-cell
S R Knezevich et al.
Cancer research, 58(22), 5046-5048 (1998-11-21)
Congenital mesoblastic nephroma (CMN) is an infantile spindle cell tumor of the kidney that is subdivided into "classical" and "cellular" forms based on the degree of cellularity and mitotic activity. The histogenesis of CMN remains obscure, but relationships to other
Rohan Kodgule et al.
Blood cancer discovery, 4(1), 34-53 (2022-11-10)
Distal enhancers play critical roles in sustaining oncogenic gene-expression programs. We identify aberrant enhancer-like activation of GGAA tandem repeats as a characteristic feature of B-cell acute lymphoblastic leukemia (B-ALL) with genetic defects of the ETV6 transcriptional repressor, including ETV6-RUNX1+ and
Hanno Hock et al.
Genes & development, 18(19), 2336-2341 (2004-09-17)
Hematopoietic stem cells (HSCs) sustain blood formation throughout life. Pathways regulating maintenance of adult HSCs are largely unknown. Here we report that the Ets-related transcription factor Tel/Etv6, the product of a locus frequently involved in translocations in leukemia, is a
S R Knezevich et al.
Nature genetics, 18(2), 184-187 (1998-02-14)
Congenital (or infantile) fibrosarcoma (CFS) is a malignant tumour of fibroblasts that occurs in patients aged two years or younger. CFS is unique among human sarcomas in that it has an excellent prognosis and very low metastatic rate. CFS is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service