Skip to Content
Merck
All Photos(2)

Documents

HPA001474

Sigma-Aldrich

Anti-OASL antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-59 kDa 2′→5′-Oligoadenylate synthetase-like protein antibody produced in rabbit, Anti-TRIP-14 antibody produced in rabbit, Anti-Thyroid receptor-interacting protein 14 antibody produced in rabbit, Anti-p59 OASL antibody produced in rabbit, Anti-p59OASL antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

EVLDAVRTVEEFLRQEHFQGKRGLDQDVRVLKVVKVGSFGNGTVLRSTREVELVAFLSCFHSFQEAAKHHKDVLRLIWKTMWQSQDLLDLGLEDLRMEQRVPDALVFTIQTRGTAEPITVTIVPAYRAL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OASL(8638)

Immunogen

59 kDa 2′→5′-Oligoadenylate synthetase-like protein recombinant protein epitope signature tag (PrEST)

Application

Anti-OASL antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

OASL (2′-5′-oligoadenylate synthetase-like) is a 59kDa protein that is an atypical member of the OAS family as it does not possess the characteristic 2′-5′ oligoadenylate synthetase activity. It exhibits an antiviral effect against the single-stranded RNA viruses, such as, picornavirus, encephalomyocarditis virus. It inhibits hepatitis C viral replication in vitro. OASL interacts with the transcriptional repressor Methyl CpG-binding protein 1 (MBD1) upon interferon stimulation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83083

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariko Ishibashi et al.
Biochemical and biophysical research communications, 392(3), 397-402 (2010-01-16)
We found the 2',5'-oligoadenylate synthetase-like (OASL) gene to be significantly elevated by high virus loads in human liver infected with hepatitis C virus (HCV). Here, we determined whether OASL inhibited HCV replication using an in vitro system. We constructed three
Joao Marques et al.
The Journal of general virology, 89(Pt 11), 2767-2772 (2008-10-22)
Viral infection of mammalian cells prompts the innate immune system to initiate an antiviral response. The recognition of the virus triggers several antiviral signalling pathways, which among others include the family of 2'-5' oligoadenylate synthetase (OAS) proteins. The p59 protein
Jesper B Andersen et al.
European journal of biochemistry, 271(3), 628-636 (2004-01-20)
The human 2'-5' oligoadenylate synthetases (OAS) form a conserved family of interferon-induced proteins consisting of four genes: OAS1, OAS2, OAS3 and the 2'-5' oligoadenylate synthetase-like gene (OASL). When activated by double-stranded RNA, OAS1-3 polymerize ATP into 2'-5'-linked oligoadenylates; 2'-5'-linked oligoadenylates
Shoichiro Tanaka et al.
Diabetes, 58(10), 2285-2291 (2009-07-31)
Fulminant type 1 diabetes is characterized by the rapid onset of severe hyperglycemia and ketoacidosis, with subsequent poor prognosis of diabetes complications. Causative mechanisms for accelerated beta-cell failure are unclear. Subjects comprised three autopsied patients who died from diabetic ketoacidosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service