H4891
Hanks′ Balanced Salts
Modified, without calcium chloride, magnesium sulfate, phenol red and sodium bicarbonate, powder, suitable for cell culture
Pharma Manufacturing
Synonym(s):
Balanced Salt Solution, Hanks’ Salt Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Formulation: click here.
Application
Hanks′ Balanced Salts may be used when working with stem cells or when growing cells at low densities. Also, it may be used to minimize cell attachment or aggregation, and for the preparation of external bathing solution and wash solution.
Quantity
Formulated to contain 9.5 grams of powder per liter of medium.
Reconstitution
Supplement with 0.35 g/L sodium bicarbonate.
Other Notes
The pH is adjusted after addition of sodium bicarbonate. Investigators who work with stem cells or cell clones frequently prefer media without phenol red.
supplement
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marta Filipiak et al.
Fish physiology and biochemistry, 38(2), 355-362 (2011-05-27)
The aim of this study was to compare DNA content in hepatocyte and erythrocyte nuclei of the European sunbleak, Leucaspius delineatus, in relation to nuclear and cell size by means of flow cytometry and fluorescence microscopy. The DNA standards, chicken
Michael C Kelly et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(19), 6699-6710 (2012-05-11)
Hearing impairment due to the loss of sensory hair cells is permanent in humans. Considerable interest targets the hair cell differentiation factor Atoh1 as a potential tool with which to promote hair cell regeneration. We generated a novel mouse model
Fc gamma receptor IIb participates in maternal IgG trafficking of human placental endothelial cells.
Tomoko Ishikawa et al.
International journal of molecular medicine, 35(5), 1273-1289 (2015-03-18)
The human placental transfer of maternal IgG is crucial for fetal and newborn immunity. Low-affinity immunoglobulin gamma Fc region receptor IIb2 (FCGR2B2 or FcγRIIb2) is exclusively expressed in an IgG-containing, vesicle-like organelle (the FCGR2B2 compartment) in human placental endothelial cells;
Ying Zhang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(18), 2001797-2001797 (2020-10-02)
In the last decade, organoid technology has developed as a primary research tool in basic biological and clinical research. The reliance on poorly defined animal-derived extracellular matrix, however, severely limits its application in regenerative and translational medicine. Here, a well-defined
Hui-Ju Yang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(13), 2828-2841 (2021-02-27)
Common fusion machinery mediates the Ca2+-dependent exocytosis of synaptic vesicles (SVs) and dense-core vesicles (DCVs). Previously, Synapsin Ia (Syn Ia) was found to localize to SVs, essential for mobilizing SVs to the plasma membrane through phosphorylation. However, whether (or how)
Protocols
Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service