Skip to Content
Merck
All Photos(1)

Documents

H4641

Sigma-Aldrich

Hanks′ Balanced Salt Solution 10x

Without calcium chloride, magnesium sulfate and sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Hanks’ Balanced Salt solution, HBSS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

form

liquid

concentration

10 ×

technique(s)

cell culture | mammalian: suitable

components

phenol red: yes

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Balanced salt solution (BSS) is crucial in cell culture as it serves as an irrigating, transporting, and diluting agent to maintain the intra- and extracellular osmotic balance and also provides energy for cell metabolism. It provides water and bulk inorganic ions to the cells to maintain the normal cell metabolism. BSS plays a role in the buffering system to maintain the medium within the physiological pH range (7.2-7.6).

Application

Hanks′ Balanced Salt Solution 10x has been used:

  • as a diluting agent during the Chinese hamster ovary cell line detachment from growth medium
  • to wash the biopsy specimens of lamina propria lymphocytes (LPLs)
  • as a wash solution to wash the fish cell line during removal from growth medium

Reconstitution

Supplement with 0.35 g/L sodium bicarbonate at 1X.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iain M Dykes et al.
Scientific reports, 8(1), 10439-10439 (2018-07-12)
ZIC2 mutation is known to cause holoprosencephaly (HPE). A subset of ZIC2 HPE probands harbour cardiovascular and visceral anomalies suggestive of laterality defects. 3D-imaging of novel mouse Zic2 mutants uncovers, in addition to HPE, laterality defects in lungs, heart, vasculature
Bastian Barker Rasmussen et al.
Applied and environmental microbiology, 82(15), 4802-4810 (2016-05-29)
Minimizing the use of antibiotics in the food production chain is essential for limiting the development and spread of antibiotic-resistant bacteria. One alternative intervention strategy is the use of probiotic bacteria, and bacteria of the marine Roseobacter clade are capable
Alison Kuchta et al.
Clinical and vaccine immunology : CVI, 18(8), 1371-1377 (2011-06-24)
Vibrio cholerae O1 is a noninvasive enteric pathogen and serves as a model for studies of mucosal immunity. Although symptomatic V. cholerae infection induces durable protection against subsequent disease, vaccination with oral killed whole-cell V. cholerae stimulates less long-lasting protection
Peimin Dai et al.
Life sciences, 127, 66-72 (2015-03-07)
Aconitum alkaloids mainly contain highly toxic aconitine (AC), mesaconitine (MA), and hypaconitine (HA) and less toxic benzoylaconine (BAC), benzoylmesaconine (BMA), benzoylhypaconine (BHA), aconine, mesaconine, and hypaconine. The efflux transporters including P-glycoprotein (P-gp), breast cancer resistance protein (BCRP) and multidrug resistance-associated
Angelika Vižintin et al.
Bioelectrochemistry (Amsterdam, Netherlands), 140, 107798-107798 (2021-03-21)
Nanosecond electric pulses have several potential advantages in electroporation-based procedures over the conventional micro- and millisecond pulses including low level of heating, reduced electrochemical reactions and reduced muscle contractions making them alluring for use in biomedicine and food industry. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service