Skip to Content
Merck
All Photos(1)

Key Documents

G5384

Sigma-Aldrich

Gelatin-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

protein from Porcine skin

Quality Level

form

saline suspension

extent of labeling

≥3 mg per mL

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

Gelatin-Agarose has been used:
  • in the isolation of exosomes from human plasma
  • to obtain fluorescent labeled enzyme For fluorescence photobleaching recovery (FPR) experiments
  • in the purifaction of matrix metalloproteinase-9 by affinity chromatography
Gelatin-agarose is an agarose in saline suspension used for affinity chromatography, protein chromatography and specialty resins. Gelatin-agarose has been used in studies informing pathology-related processes of tissue remodeling as well as sperm-egg interactions in mammals.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R P Kitson et al.
Journal of immunology (Baltimore, Md. : 1950), 160(9), 4248-4253 (1998-05-09)
We have previously documented that adoptively transferred IL-2-activated NK (A-NK) cells can accumulate within cancer metastases. Electron microscopic studies of pulmonary metastases have revealed that adoptively transferred A-NK cells that accumulate within metastases bind to endothelial cells and are able
S A Corbett et al.
Blood, 88(1), 158-166 (1996-07-01)
Plasma fibronectin (pFN) cross-linked to fibrin during the injury response provides a provisional matrix required for cells to begin tissue repair. Using a synthetic matrix of pFN and fibrin as a substrate for cell adhesion and spreading, we have determined
Annick Bergeron et al.
Molecular reproduction and development, 71(4), 461-470 (2005-05-14)
Mammalian seminal plasma contains among others, two major families of proteins, namely spermadhesins and those proteins that contain fibronectin type II domains. Spermadhesins are the major proteins of boar and stallion seminal plasma and homologous proteins have been identified in
Mélanie Boisvert et al.
Biology of reproduction, 70(3), 656-661 (2003-10-31)
Bovine seminal plasma (BSP) contains a family of major proteins designated BSP-A1/A2, BSP-A3, and BSP-30kDa (collectively called BSP proteins) that bind to sperm at ejaculation and potentiate sperm capacitation. Homologous proteins have been identified in stallion, boar, goat, and ram
G A Piazza et al.
The Biochemical journal, 262(1), 327-334 (1989-08-15)
Dipeptidyl peptidase IV (DPP IV) is a cell surface glycoprotein which has been implicated in hepatocyte-extracellular matrix interactions [Hixson, DeLourdes, Ponce, Allison & Walborg (1984) Exp. Cell Res. 152, 402-414; Walborg, Tsuchida, Weeden, Thomas, Barrick, McEntire, Allison & Hixson (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service