Skip to Content
Merck

Skip To

H8280

N-Hydroxysuccinimidyl-Sepharose 4 Fast Flow

matrix fast-flow highly cross-linked 4% beaded agarose

Synonym(s):

N-Hydroxysuccinimidyl-Agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
23151817
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

isopropanol suspension

Quality Segment

extent of labeling

16-23 μmol per mL

technique(s)

protein array: suitable

matrix

fast-flow highly cross-linked 4% beaded agarose

matrix active group

N-hydroxysuccinimide active ester

matrix spacer

6 atoms (when ligands are coupled by displacement of NHS)

storage temp.

2-8°C

General description

N-Hydroxysuccinimidyl-Sepharose 4 Fast Flow is a cross-linked and preactivated matrix obtained by mixing Sepharose 4 Fast Flow with 6-aminohexanoic acid through a spacer arm. Esterification with N-hydroxysuccinimide activates the terminal carboxyl group. Ligand-containing primary amino groups directly interact with this active ester to form a chemically very stable amide linkage.

Application

N-Hydroxysuccinimidyl-Sepharose 4 Fast Flow has been used as a part of a magnetic particles-based visual detection system to achieve separation upon binding of the amplified gene. It has also been used to immobilize protein during pull-down assay.

Features and Benefits

  • Specifically recommended for coupling of small amino-containing proteins and peptides in process-scale applications
  • High level of pre-activation, which gives a high degree of substitution of the selected ligand
  • NHS coupling method enables chemically stable ligand attachment

Legal Information

Sepharose is a trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C5338GE17-0906-01P7892
matrix active group

N-hydroxysuccinimide active ester

matrix active group

cyanate or related structures

matrix active group

N-hydroxysuccinimide (NHS) activated

matrix active group

-

matrix

fast-flow highly cross-linked 4% beaded agarose

matrix

Sepharose 4 Fast Flow

matrix

4% cross-linked agarose

matrix

agarose, 4% cross-linked

form

isopropanol suspension

form

lyophilized powder

form

resin

form

suspension

technique(s)

protein array: suitable

technique(s)

affinity chromatography: suitable

technique(s)

liquid chromatography (LC): suitable

technique(s)

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

53.6 °F

flash_point_c

12 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters