This material is the pure (-)-Naloxone. The product is supplied by the British Pharmacopoeia. A link to the supplier documentation is provided below:
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
API family
naloxone
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
[Cl-].[N+H]1([C@H]2[C@]3([C@@]4([C@@H](Oc5c4c(ccc5O)C2)C(=O)CC3)CC1)O)CC=C
InChI
1S/C19H21NO4.ClH/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11;/h2-4,14,17,21,23H,1,5-10H2;1H/t14-,17+,18+,19-;/m1./s1
InChI key
RGPDIGOSVORSAK-STHHAXOLSA-N
General description
Application
Also used in monographs such as:
Packaging
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename BP | manufacturer/tradename EDQM | manufacturer/tradename BP | manufacturer/tradename BP |
| format neat | format neat | format neat | format neat |
| shelf life limited shelf life, expiry date on the label | shelf life - | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
| form solid | form - | form solid | form solid |
| application(s) pharmaceutical | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) pharmaceutical |
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| BP548 | 04061835278169 |
-
Is this a racemic mix or pure (-)-Naloxone?
1 answer-
Helpful?
-



