Skip to Content
Merck
All Photos(1)

Key Documents

F9665

Sigma-Aldrich

Fetal Bovine Serum

Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

HI FBS, HI FCS, HI Fetal Calf Serum, HI Serum, HI sera, Heat Inactivated Fetal Calf Serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71
biological source:
bovine fetus
origin:
non-USA origin
sterility:
sterile-filtered
shipped in:
dry ice

biological source

bovine fetus

Quality Level

sterility

sterile-filtered

product line

FBS Classic

composition

Hemoglobin, ≤25 mg/dL

origin

non-USA origin

availability

not available in USA

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

FBS is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.

Preparation Note

Prepared by heating at 56°C for 30 minutes. Heating serum to 56°C for 30 minutes is used to inactivate the complement system for immunoassays. Heat inactivation also inactivates other undetermined inhibitors of cell growth in culture.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Legal Information

Not available in USA or Canada. Please check with your local supplier regarding availability and import restrictions in your country.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Karoline Pill et al.
Frontiers in bioengineering and biotechnology, 6, 156-156 (2018-11-10)
A promising approach to overcome hypoxic conditions in tissue engineered constructs is to use the potential of endothelial cells (EC) to form networks in vitro when co-cultured with a supporting cell type in a 3D environment. Adipose tissue-derived stromal cells
Athar Khalil et al.
Frontiers in oncology, 8, 482-482 (2018-11-15)
T-box (TBX) transcription factors are evolutionary conserved genes and master transcriptional regulators. In mammals, TBX2 subfamily (TBX2, TBX3, TBX4, and TBX5) genes are expressed in the developing lung bud and tracheae. Our group previously showed that the expression of TBX2
Jing Lin et al.
Nature communications, 10(1), 3544-3544 (2019-08-09)
Simultaneous measurement of proteins and mRNA in single cells enables quantitative understanding and modeling of cellular functions. Here, we present an automated microfluidic system for multi-parameter and ultra-sensitive protein/mRNA measurements in single cells. Our technology improves the sensitivity of digital
Amina Kamar et al.
Frontiers in genetics, 8, 217-217 (2018-01-13)
Despite an obvious role for consanguinity in congenital heart disease (CHD), most studies fail to document a monogenic model of inheritance except for few cases. We hereby describe a first-degree cousins consanguineous Lebanese family with 7 conceived children: 2 died
Xiaolu Zhang et al.
Nature communications, 10(1), 4732-4732 (2019-10-20)
Current multiplexing strategies for massively parallel sequencing of genomic DNA mainly rely on library indexing in the final steps of library preparation. This procedure is costly and time-consuming, because a library must be generated separately for each sample. Furthermore, library

Articles

Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.

Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.

Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.

Review the basics of using fetal bovine (or calf) serum for cell culture and how sourcing and regulations affect the price of sera for cell-based assays.

Related Content

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service