Skip to Content
Merck
All Photos(1)

Key Documents

P0781

Sigma-Aldrich

Penicillin-Streptomycin

liquid, non-animal origin, suitable for cell culture, BioReagent

Synonym(s):

Pen-Strep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.76

Product Name

Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

concentration

100 ×

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes
protein synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(O)=O.CNC[C@H]4[C@H](O)[C@@H](O)[C@H](CCO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)C5(O)C=O)O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(N)=N)[C@@H](O)[C@@H]6NC(N)=N

InChI

1S/C23H43N7O12.C16H18N2O4S/c1-7-23(38,6-32)18(42-19-8(5-28-2)12(33)13(34)9(40-19)3-4-31)20(39-7)41-17-11(30-22(26)27)14(35)10(29-21(24)25)15(36)16(17)37;1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9/h6-20,28,31,33-38H,3-5H2,1-2H3,(H4,24,25,29)(H4,26,27,30);3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t7-,8-,9-,10+,11-,12-,13-,14+,15-,16+,17+,18-,19-,20-,23+;11-,12+,14-/m01/s1

InChI key

KMOOCZWLFBSQCW-XFOLRDBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Penicillin streptomycin, also known as Pen-Strep, is the most commonly used antibiotic solution for culture of mammalian cells. Penicillin Streptomycin solution is used to maintain sterile conditions during cell culture, and they show few adverse effects on the cultured cells.

Application

Penicillin-Streptomycin solution is used to supplement cell culture media to control bacterial contamination. It has also been used for the preparation of lipid emulsions. It is recommended for use in cell culture applications at 10 mL/L.

Components

This product is formulated to contain 10,000 units penicillin and 10 mg streptomycin/mL.

Other Notes

20x100ml,20ml,100ml
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan Brown et al.
Tissue engineering, 12(3), 519-526 (2006-04-04)
The extracellular matrix (ECM) has been successfully used as a scaffold for constructive remodeling of multiple tissues in both preclinical studies and in human clinical applications. The basement membrane is a specialized form of the ECM that supports and facilitates
D T Stein et al.
The Journal of clinical investigation, 100(2), 398-403 (1997-07-15)
Lowering of the elevated plasma FFA concentration in 18- 24-h fasted rats with nicotinic acid (NA) caused complete ablation of subsequent glucose-stimulated insulin secretion (GSIS). Although the effect of NA was reversed when the fasting level of total FFA was
Tinh-Hai Collet et al.
Molecular metabolism, 6(10), 1321-1329 (2017-10-17)
Pro-opiomelanocortin (POMC)-derived peptides act on neurons expressing the Melanocortin 4 receptor (MC4R) to reduce body weight. Setmelanotide is a highly potent MC4R agonist that leads to weight loss in diet-induced obese animals and in obese individuals with complete POMC deficiency.
Zijin Zhou et al.
International journal of food microbiology, 257, 176-182 (2017-07-03)
The efficacy of levulinic acid (LVA) in combination with sodium dodecyl sulfate (SDS) in removal of foodborne viruses, enteric bacterial pathogens and their surrogates on fresh strawberries was investigated. Inoculated strawberries were treated with potable water, sodium hypochlorite solution (50ppm)
184: Toxic effects of antibiotics routinely used in cell culture on primary cultures of dissociated mouse brain cells.
Amonn F
Pediatric Research, 10(10), 900-900 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service