Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Fluorouracil
Synonym(s):
2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1
H
,3
H
)-pyrimidinedione, 5-Fluorouracil
Empirical Formula (Hill Notation):
C
4
H
3
FN
2
O
2
CAS Number:
51-21-8
Molecular Weight:
130.08
EC Number:
200-085-6
Products
Product Category
bioactive small molecules (1)
fluorescence reagent (1)
Brand
Sigma-Aldrich (3)
Supelco (1)
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
F6627
5-Fluorouracil,
≥99% purity (HPLC), powder
Expand
View Pricing
343922
5-Fluorouracil,
≥98% (HPLC), solid, antitumor agent, Calbiochem
®
Expand
View Pricing
8.18505
5-Fluorouracil,
for synthesis
Expand
View Pricing
03738
5-Fluorouracil,
analytical standard
Expand
View Pricing