Skip to Content
Merck

Skip To

F0635

Rat Fibronectin

from rat plasma, powder, suitable for cell culture

Synonym(s):

Fibronectin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
eCl@ss:
42020141
NACRES:
NA.75
EC Number:
289-149-2
MDL number:
Biological source:
rat plasma
Form:
powder
Technique(s):
cell culture | mammalian: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fibronectin rat plasma, powder, BioReagent, suitable for cell culture

biological source

rat plasma

Quality Segment

sterility

sterile

product line

BioReagent

form

powder

packaging

pkg of 0.5 mg, pkg of 1 mg, pkg of 2 mg

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑5 μg/cm2

solubility

H2O: 1 mg/mL at 37 °C, clear, colorless (some particles)

UniProt accession no.

binding specificity

Peptide Source: Collagen

shipped in

dry ice

storage temp.

−20°C

Gene Information

rat ... Fn1(25661)

General description

Fibronectin (FN)/FN1 (fibronectin 1) is a multifunctional glycoprotein, produced by hepatic stellate cells (HSCs). It is a constituent of extracellular matrix (ECM). FN1 is found in the cell membrane and cytoplasm. It is also synthesized and liberated by fibroblasts, endothelial cells, macrophages and hepatocytes in vivo.

Application

Fibronectin rat plasma has been used in the induction of differentiation , adhesion to extracellular matrix, neural differentiation [1] and primary neuronal ventral mesencephalon cultures.

Biochem/physiol Actions

Fibronectin (FN)/FN1 (fibronectin 1) plays a major role in cell cycle progression, cell adhesion and proliferation and fibrotic progression. It is essential for for collagen matrix assembly. High expression of FN has a vital role during liver fibrosis. FN1 helps in the support of other matrix proteins.

Preparation Note

This product is lyophilized from 0.05 M Tris buffered saline at pH 7.5. It is soluble at 1 mg/mL in water, and should be allowed to dissolve for at least 30 minutes at 37°C. A small amount of undissolved material may remain, but it will not affect performance. The protein homogeneity is evaluated by immunoelectrophoresis, showing a single arc, is endotoxin tested, and the source material was tested negative for HIV, hepatitis B and hepatitis C.

In coating culture surfaces, fibronectin should be diluted in sterile balanced sterile salt solution and coated with minimal volume. Surface should then be air dried for 45 minutes at room temp and can be stored for 2-4 weeks at 2-8°C.

Analysis Note

A single arc is shown when reacted with anti-fibronectin by IEP (immunoelectrophoresis).

Other Notes

Fibronectin is an adhesive glycoproteins found in both the cell surface and in plasma.

Disclaimer

The reconstituted solution should be stored in working aliquots at -20°C or lower. Vortexing, excessive agitation, or additoinal freezing and thawing of reconstituted fibronectin are not recommended.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F1056F4759F0162
sterility

sterile

sterility

sterile

sterility

sterile

sterility

-

biological source

rat plasma

biological source

human plasma

biological source

bovine plasma

biological source

human plasma

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

lyophilized powder

form

powder

form

lyophilized powder

packaging

pkg of 0.5 mg, pkg of 2 mg, pkg of 1 mg

packaging

pkg of 1 mg, pkg of 2 mg, pkg of 5 mg

packaging

pkg of 1 mg, pkg of 2 mg, pkg of 5 mg

packaging

pkg of 0.5 mg


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters