Skip to Content
Merck
All Photos(1)

Key Documents

E7034

Sigma-Aldrich

EX-527

≥98% (HPLC)

Synonym(s):

6-Chloro-2,3,4,9-tetrahydro-1H-Carbazole-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13ClN2O
CAS Number:
Molecular Weight:
248.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

NC(=O)C1CCCc2c1[nH]c3ccc(Cl)cc23

InChI

1S/C13H13ClN2O/c14-7-4-5-11-10(6-7)8-2-1-3-9(13(15)17)12(8)16-11/h4-6,9,16H,1-3H2,(H2,15,17)

InChI key

FUZYTVDVLBBXDL-UHFFFAOYSA-N

Application

EX-527 has been used:
  • in 1% dimethyl sulfoxide, 30%, polyethylene glycol-400 and 1% Tween 80 for treating C57BL/6 N mice to study its effect on intestinal morphological changes and crypt cell apoptosis
  • as a an inhibitor of sirtuin 1, in treating human cancer lines MCF-7 (Michigan cancer foundation-7) and HCT116 (colon cancer cell line) incubated in Dulbecco′s modified Eagle′s medium, to study its effect on mitochondrial ATP (adenosine triphosphate) production
  • Intracerebroventricularly infused in rat model of epileptogenesis, to access kainic acid–induced status epilepticus stimulated sirtuin 1 activity

Biochem/physiol Actions

EX-527 is a potent and selective sirtuin 1 (SIRT1) inhibitor (IC50 38 nM) identified from a high throughput screen. EX-527 is more selective (200-500-fold) for SIRT1 than for SIRT2 or SIRT3 and has been shown to be a potent SIRT6 inhibitor using H3K56 deacetylation site based substrate. EX-527 does not inhibit class I/II HDAC activity at concentrations up to 100uM. Enhances p53 acetylation in response to DNA damaging agents. EX-527 is racemic; the active isomer (EX-243) gives similar results and potency whereas the other isomer (designated EX-242) is inactive.
Potent SIRT1 and SIRT6 inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Rise in ATP, ROS, and Mitochondrial Content upon Glucose Withdrawal Correlates with a Dysregulated Mitochondria Turnover Mediated by the Activation of the Protein Deacetylase SIRT1
Song S and Hwang E
Cells, 8(1), 11-11 (2019)
Hong-Xia Liu et al.
Experimental cell research, 357(2), 271-281 (2017-05-30)
Mitochondrial trifunctional protein α-subunit (MTPα) is involved in the fatty acid β-oxidation (FAO) pathway. Two MTPα activities, 3-hydroxyacyl-CoA dehydrogenase and long-chain hydratase, have been linked with the occurrence and development of obesity and obesity-related disorders. These activities catalyze two steps
Shuangdong Chen et al.
Biochemical and biophysical research communications, 516(4), 1196-1203 (2019-07-13)
Sirtuin1 (SIRT1), which is regulated by microRNA-34a (miR-34a), can modulate pathophysiology processes, including nonalcoholic fatty liver disease and intestinal ischemia/reperfusion injury. We previously reported that SIRT1, an NAD+-dependent deacetylase, plays a vital role in the development of neuropathic pain. However
Ayana N Martin et al.
Journal of immunology research, 2018, 2402593-2402593 (2018-08-03)
Resistance and tolerance to infection are two universal fitness and survival strategies used by inflammation and immunity in organisms and cells to guard homeostasis. During sepsis, however, both strategies fail, and animal and human victims often die from combined innate
The Role of Sirt1 in Epileptogenesis
Hall Alicia M, et al.
eNeuro, 4(1) (2017)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service