Skip to Content
Merck

566313

SIRT1 Activator II

The SIRT1 Activator II, also referenced under CAS 374922-43-7, modulates the biological activity of SIRT1. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

SIRT1 Activator II, 3-(Benzenesulfonyl)-1-(4-fluorophenyl)pyrrolo[4,5-b]quinoxalin-2-amine , SIRT1 Activator II, CAY10602

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H15FN4O2S
CAS Number:
Molecular Weight:
418.44
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

FC(C=C1)=CC=C1N2C(N)=C(S(C3=CC=CC=C3)(=O)=O)C4=C2N=C5C(C=CC=C5)=N4

InChI

1S/C22H15FN4O2S/c23-14-10-12-15(13-11-14)27-21(24)20(30(28,29)16-6-2-1-3-7-16)19-22(27)26-18-9-5-4-8-17(18)25-19/h1-13H,24H2

InChI key

CSFVFDHRYKBBPD-UHFFFAOYSA-N

General description

A cell-permeable pyrroloquinoxaline compound that acts as a reversible SIRT1 activator (10 µM causes ~2.3-fold fluorescent enhancement in a fluorescent assay using hrSIRT1) and enhances fat mobilization in fully differentiated 3T3L1 fibroblasts. Shown to inhibit LPS-induced TNF-α release in THP-1 cells by ~10-fold more potent than resveratrol (Cat. No. 554325).

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lavu, S., et al. 2008. Nat. Rev. Drug Discov.7, 841.
Nayagam, V.M., et al. 2006. J. Biomol. Screen.11, 959.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library