Skip to Content
Merck
All Photos(1)

Key Documents

DNASE70

Sigma-Aldrich

On-Column DNase I Digestion Set

sufficient for 70 preparations

Synonym(s):

Deoxyribonuclease I Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.52

grade

Molecular Biology
for molecular biology

form

liquid

usage

sufficient for 70 preparations

technique(s)

RNA purification: suitable

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The On-Column DNase I Digestion Set offers a simple and convenient method for eliminating genomic DNA from total RNA preparations. This Set is intended for use with the GenElute Mammalian Total RNA Miniprep Kit and the Spectrum Plant Total RNA Kit. It can also be used with many other commercially available RNA purification kits.
The vast majority of DNA is eliminated from total RNA purifications by the procedures provided in the standard protocols of the Sigma Kits named below. However, for very sensitive applications, such as quantitative RT-PCR, even trace amounts of DNA can result in false positive detection of cDNA-derived amplicons.

The On-Column DNase I Digestion Set offers a simple and convenient method for eliminating genomic DNA from total RNA preparations. This Set is intended for use with the GenElute Mammalian Total RNA Miniprep Kit, the GenElute Bacterial Total RNA Purification Kit, the GenElute Yeast Total RNA Purification Kit and the Spectrum Plant Total RNA Kit. However, the digestion protocol is compatible with many other commercially available RNA purification kits.

The term ′On-Column′ indicates that digestion is accomplished in-process for true convenience. Each of Sigma′s total RNA purification kits utilize a binding column for purification. DNase I digestion is accomplished in 15 minutes while total RNA is bound to the column substrate. Several wash steps and elution of the total RNA immediately follow. To review the complete protocol, see the Technical Bulletin for any of the Kits named above.

Application

On-Column DNase I Digestion Set is suitable for removing gDNA from total RNA preparations. It may not be suitable for RT-PCR applications as a small amount of residual DNA may remain. On-Column DNase I Digestion Set was used to purify RNA preparations isolated from Vitis vinifera cultivars.

Features and Benefits

The On-Column DNase I Digestion Set offers a simple and convenient method for eliminating genomic DNA from total RNA preparations. This Set is intended for use with the GenElute Mammalian Total RNA Miniprep Kit, the GenElute Bacterial Total RNA Purification Kit, the GenElute Yeast Total RNA Purification Kit and the Spectrum Plant Total RNA Kit. However, the digestion protocol is compatible with many other commercially available RNA purification kits.

Principle

The on-column digestion is accomplished in-process for convenience. DNase I digestion is accomplished in 15 minutes while total RNA is bound to the column substrate. Several wash steps and elution of the total RNA immediately follow.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
Spectrum is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filipa Monteiro et al.
PloS one, 8(9), e72998-e72998 (2013-09-12)
The pivotal role of cultivated grapevine (Vitis vinifera L.) in many countries economy is compromised by its high susceptibility to Plasmopara viticola, the causal agent of downy mildew disease. Recent research has identified a set of genes related to resistance
Hubert Sytykiewicz et al.
International journal of molecular sciences, 21(17) (2020-09-04)
Thioredoxins (Trxs) and thioredoxin reductases (TrxRs) encompass a highly complex network involved in sustaining thiol-based redox homeostasis in plant tissues. The purpose of the study was to gain a new insight into transcriptional reprogramming of the several genes involved in
Carmen Michán et al.
FEMS yeast research, 9(7), 1078-1090 (2009-08-26)
We report the absolute transcription profiles of 24 genes coding for putative thioredoxin (Trx)- and glutathione (GSH)-dependent redox system components, accompanying the Candida albicans batch culture growth, under either yeast or hyphal conditions. All mRNAs investigated (plus the housekeeping ACT1)
Alexander V Gopanenko et al.
Cells, 9(5) (2020-05-21)
An imbalance in the synthesis of ribosomal proteins can lead to the disruption of various cellular processes. For mammalian cells, it has been shown that the level of the eukaryote-specific ribosomal protein eL29, also known as the one interacting with
Ghazanfar A Chishti et al.
Animals : an open access journal from MDPI, 10(4) (2020-04-25)
Transition to grain increases inflammation and causes parakeratosis, which can decrease growth performance in fattening animals. It is unknown if ruminants adapt to these inflammatory responses over time. In a three-phase, 49-day experiment, all wethers (n = 13, BW =

Related Content

The Spectrum Plant Total RNA Kit incorporates a new, patent-pending lysis and binding chemistry. This kit is intended to purify total RNA from species and tissue where most Total RNA protocols fail. Without sacrificing affordability, you can purify more total RNA per isolation.

GenElute™-E Single Spin DNA and RNA purification technology enables rapid DNA and RNA extraction and purification from mammalian cells, blood, tissue, and plant tissue samples.

GenElute™-E Single Spin DNA and RNA purification technology enables rapid DNA and RNA extraction and purification from mammalian cells, blood, tissue, and plant tissue samples.

GenElute™-E Single Spin DNA and RNA purification technology enables rapid DNA and RNA extraction and purification from mammalian cells, blood, tissue, and plant tissue samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service