Skip to Content
Merck
All Photos(2)

Documents

D7071

Sigma-Aldrich

Dapoxetine hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-N,N-dimethyl-1-phenyl-3-(1-naphthalenyloxy)propanamine hydrochloride, S-(+)-N,N-dimethyl-a-[2-(naphthalenyloxy)ethyl] benzenemethanamine hydrochloride, LY-210448 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO · HCl
CAS Number:
Molecular Weight:
341.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +125 to +135°, c = 1 in methanol

storage condition

desiccated

color

white

solubility

DMSO: ≥20 mg/mL

originator

Eli Lilly

storage temp.

room temp

SMILES string

Cl.CN(C)[C@@H](CCOc1cccc2ccccc12)c3ccccc3

InChI

1S/C21H23NO.ClH/c1-22(2)20(18-10-4-3-5-11-18)15-16-23-21-14-8-12-17-9-6-7-13-19(17)21;/h3-14,20H,15-16H2,1-2H3;1H/t20-;/m0./s1

InChI key

IHWDIQRWYNMKFM-BDQAORGHSA-N

Gene Information

human ... SLC6A4(6532)

General description

Dapoxetine ((+)-(S)-N,N-dimethyl-(α)-[2(1naphthal enyloxy)ethyl]-benzenemethanamine hydrochloride) possess a similar structure as that of fluoxetine.

Biochem/physiol Actions

Dapoxetine is capable of blocking recombinant Kv4.3 potassium voltage-gated channels. It is considered as safe.
Potent Selective serotonin reuptake inhibitor (SSRI); used in treatment of premature ejaculation

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Clément et al.
The journal of sexual medicine, 9(10), 2562-2573 (2012-08-22)
A brain network specifically activated when ejaculation occurs has been described in rats. Increasing serotonin (5-hydroxytryptamine [5-HT]) tone impairs ejaculation and chronic 5-HT selective serotonin reuptake inhibitors (SSRIs) are known to inhibit ejaculation. However, efficacy of acute treatment with SSRI
Tae Kon Kim et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 926, 42-46 (2013-04-02)
In this study, we describe and validate a rapid and sensitive method for quantitation of dapoxetine in rat plasma by using ultra-performance liquid chromatography-electrospray ionization tandem mass spectrometry (UPLC-ESI/MS/MS). Plasma samples were prepared by protein precipitation with acetonitrile, and sildenafil
Stanley E Althof et al.
The journal of sexual medicine, 7(6), 2243-2252 (2010-04-07)
The Clinical Global Impression of Change (CGIC) measures have high utility in clinical practice. However, it is unknown whether the CGIC is valued for assessing premature ejaculation (PE) symptoms and/or the relationship between CGIC and other validated PE patient-reported measures.
Dapoxetine: a new option in the medical management of premature ejaculation
McMahon CG
Therapeutic Advances in Urology, 4(5), 233-251 (2012)
Hartmut Porst et al.
The journal of sexual medicine, 7(6), 2231-2242 (2010-04-24)
Premature ejaculation (PE) is classified as an acquired or lifelong condition but data on baseline characteristics and response to treatment of men with acquired or lifelong PE and mild erectile dysfunction (ED) or normal erectile function (EF) is limited. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service