Skip to Content
Merck
All Photos(2)

Key Documents

C6072

Sigma-Aldrich

Cholesteryl palmitate

≥98% (HPLC; detection at 205 nm)

Synonym(s):

5-Cholestene 3-palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H76O2
CAS Number:
Molecular Weight:
625.06
Beilstein:
2342867
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC; detection at 205 nm)

form

powder

mp

74-77 °C (lit.)

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)O[C@H]1CC[C@]2(C)C3CC[C@]4(C)C(CCC4C3CC=C2C1)[C@H](C)CCCC(C)C

InChI

1S/C43H76O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-23-41(44)45-36-28-30-42(5)35(32-36)24-25-37-39-27-26-38(34(4)22-20-21-33(2)3)43(39,6)31-29-40(37)42/h24,33-34,36-40H,7-23,25-32H2,1-6H3/t34-,36+,37+,38-,39+,40+,42+,43-/m1/s1

InChI key

BBJQPKLGPMQWBU-JADYGXMDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cholesteryl palmitate was used as standard in HPLC analysis of lipids from cooked ground beef and rat liver and mouse muscle tissue samples.

Biochem/physiol Actions

Cholesteryl palmitate was used as standard in HPLC analysis of lipids from cooked ground beef and rat liver and mouse muscle tissue samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TBA Values and 7?Ketocholesterol in Refrigerated Raw and Cooked Ground Beef
Vore VR
Journal of Food Science, 53, 1058-1061 (1988)
K Owens et al.
Journal of lipid research, 11(5), 486-495 (1970-09-01)
Myofibrillar, mitochondrial, and microsomal fractions were prepared from normal and dystrophic mouse limb muscle by differential centrifugation and analyzed for phospholipids and cholesterol. Fatty acids and aldehydes of neutral lipids and of phospholipids from whole muscle and particulate fractions were
Karin M Thelen et al.
Journal of clinical pharmacology, 46(7), 812-816 (2006-07-01)
Statins inhibit endogenous cholesterol synthesis, up-regulate low-density lipoprotein (LDL) receptor expression in mammalian liver cells, and thus decrease circulating LDL-cholesterol concentrations. As cholesterol seems to play a role in the development of neurodegenerative diseases, it is of interest to evaluate
7-Ketocholesterol. Its effects on hepatic cholesterogenesis and its hepatic metabolism in vivo and in vitro.
S K Erickson et al.
The Journal of biological chemistry, 252(15), 5186-5193 (1977-08-10)
M Rosseneu et al.
Atherosclerosis, 108(2), 127-136 (1994-08-01)
The European Atherosclerosis Research Study (EARS) is a study of the biological expression of a paternal history of premature coronary heart disease. The study was carried out in 14 centres in 11 European countries, where the offspring of fathers who

Articles

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Cholesterol esterification enhances transport efficiency in lipoproteins for increased blood stream transport.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service