Skip to Content
Merck
All Photos(3)

Documents

D3126

Sigma-Aldrich

2-Deoxyribose 5-phosphate sodium salt

≥95% (ICP-AES)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11O7P · xNa+
CAS Number:
Molecular Weight:
214.11 (free acid basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (inorganic)

Quality Level

Assay

≥95% (ICP-AES)

form

powder

color

white to off-white

solubility

H2O: 100 mg/mL, clear, colorless to very faintly yellow

cation traces

Na: 14.0-19.0% (dry basis)

storage temp.

−20°C

SMILES string

[Na].OC(COP(O)(O)=O)C(O)CC=O

InChI

1S/C5H11O7P.Na.H/c6-2-1-4(7)5(8)3-12-13(9,10)11;;/h2,4-5,7-8H,1,3H2,(H2,9,10,11);;

InChI key

VKWGIGKPYXQSGV-UHFFFAOYSA-N

Application

2-Deoxyribose 5-phosphate is used as a substrate to identify, differentiate and characterize deoxyribose-phosphate aldolase(s). It is used for the synthesis of 2′-deoxyribonucleoside building blocks for the synthesis of antisense drugs and antiviral nucleosides.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anming Wang et al.
Bioprocess and biosystems engineering, 35(5), 857-863 (2012-02-10)
To improve the properties of the immobilised 2-deoxy-D-ribose-5-phosphate aldolase (DERA), unreacted functional groups on support surface were blocked with amino acids. The relative activities of the immobilised enzyme were 144.7 and 141.9% when the post-immobilisation modification was done with Arg
Sanni Voutilainen et al.
Applied microbiology and biotechnology, 104(24), 10515-10529 (2020-11-05)
In this work, deoxyribose-5-phosphate aldolase (Ec DERA, EC 4.1.2.4) from Escherichia coli was chosen as the protein engineering target for improving the substrate preference towards smaller, non-phosphorylated aldehyde donor substrates, in particular towards acetaldehyde. The initial broad set of mutations
Zhong-Yu You et al.
Journal of industrial microbiology & biotechnology, 40(1), 29-39 (2012-11-28)
A codon-optimized 2-deoxyribose-5-phosphate aldolase (DERA) gene was newly synthesized and expressed in Escherichia coli to investigate its biochemical properties and applications in synthesis of statin intermediates. The expressed DERA was purified and characterized using 2-deoxyribose-5-phosphate as the substrate. The specific
Yong-Mo Kim et al.
Protein expression and purification, 68(2), 196-200 (2009-06-10)
A new deoC gene encoding deoxyribose 5-phosphate aldolase (DERA) was identified in Yersinia sp. EA015 isolated from soil. The DERA gene had an open reading frame (ORF) of 672 base pairs encoding 223 amino acids to yield a protein of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service