Skip to Content
Merck
All Photos(1)

Documents

15732

Sigma-Aldrich

D-Xylulose 5-phosphate lithium salt

≥90% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11O8P · xLi+
CAS Number:
Molecular Weight:
230.11 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90% (TLC)

form

powder or crystals

storage temp.

−20°C

SMILES string

OCC([C@@H](O)[C@H](O)COP(O)(O)=O)=O

InChI

1S/C5H11O8P/c6-1-3(7)5(9)4(8)2-13-14(10,11)12/h4-6,8-9H,1-2H2,(H2,10,11,12)/t4-,5-/m1/s1

InChI key

FNZLKVNUWIIPSJ-RFZPGFLSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D-Xylulose 5-phosphate (D-Xu-5-P) is a metabolite of the hexose monophosphate pathway activating protein phosphatase 2A to mediate the acute effects of carbohydrate feeding on the glycolytic pathway, the coordinated control of enzymes required in fatty acid & triglyceride synthesis. D-Xylulose 5-phosphate is the signal for the coordinated control of lipogenesis. The elevation of D-Xylulose 5-phosphate is the coordinating signal that both acutely activates phosphofructokinase in glycolysis and promotes the action of the transcription factor ChREBP to increase transcription of the genes for the enzymes of lipogenesis, the hexose monophosphate shunt, and glycolysis, all of which are required for the de novo synthesis of fat.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Susen Becker et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 883-884, 68-75 (2011-11-15)
The analysis of metabolites in human body fluids remains a challenge because of their chemical diversity and dynamic concentration range. Liquid chromatography (LC) in combination with tandem mass spectrometry (MS/MS) offers a robust, reliable, and economical methodology for quantitative single
J J Wang et al.
Alcoholism, clinical and experimental research, 21(4), 576-580 (1997-06-01)
Thiamine deficiency, a frequent complication of alcoholism, contributes significantly to the development of damage in various organ systems, including the brain. The molecular mechanisms that underlie the differential vulnerabilities to thiamine deficiency of tissue and cell types and among individuals
Sigrun Langbein et al.
International journal of cancer, 122(11), 2422-2428 (2008-02-28)
Targeted therapies have demonstrated clinical benefit with limited impact on long-term disease specific survival in the treatment of renal cell cancer (RCC). New opportunities for the treatment of tumors that are resistant or have relapsed, are needed. Increased anaerobic glucose
Kostas Tsintzas et al.
Clinical science (London, England : 1979), 124(11), 675-684 (2013-01-16)
Physiological hyperglycaemia and hyperinsulinaemia are strong modulators of gene expression, which underpins some of their well-known effects on insulin action and energy metabolism. The aim of the present study was to examine whether acute in vivo exposure of healthy humans
S Langbein et al.
British journal of cancer, 94(4), 578-585 (2006-02-09)
Tumours ferment glucose to lactate even in the presence of oxygen (aerobic glycolysis; Warburg effect). The pentose phosphate pathway (PPP) allows glucose conversion to ribose for nucleic acid synthesis and glucose degradation to lactate. The nonoxidative part of the PPP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service