Skip to Content
Merck
All Photos(1)

Key Documents

CLS3795

Corning® 96 Well Clear Polystyrene Microplate

round bottom clear, polystyrene, bag of 25, sterile, lid: no

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear wells
polystyrene
round bottom clear
round wells
size:
96 wells

material

clear wells
polystyrene
round bottom clear
round wells

sterility

sterile

feature

lid: no
skirt

packaging

bag of 25

manufacturer/tradename

Corning 3795

size

96 wells

well volume

330 μL

well working volume

75-200 μL

color

clear

suitability

suitable for (cell cuture)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Clear Polystyrene Microplate round bottom without lid is hydrophobic and binds biomolecules through passive interactions.
  • Ideal for immobilization of large molecules, such as antibodies.
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm².
  • Conforms to standard microplate footprint and dimensions.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of
A Sundqvist et al.
Oncogene, 32(31), 3606-3615 (2012-08-29)
Deregulation of the transforming growth factor β (TGFβ) signal transduction cascade is functionally linked to cancer. In early phases, TGFβ acts as a tumor suppressor by inhibiting tumor cell proliferation, whereas in late phases, it can act as a tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service