Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
96 Well Clear Polystyrene Microplate
Products
Product Category
plates (11)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS3370
Corning
®
96 Well Clear Polystyrene Microplate,
flat bottom clear, polystyrene, sterile, lid, case of 100, pack of 20
Expand
View Pricing
CLS3788
Corning
®
96 Well Clear Polystyrene Microplate,
round bottom clear, polystyrene, bag of 20 ×, sterile, lid
Expand
View Pricing
CLS3367
Corning
®
96 Well Clear Polystyrene Microplate,
round bottom clear, polystyrene, pkg of Individually wrapped, sterile, lid: no
Expand
View Pricing
CLS3897
Corning
®
96 Well Clear Polystyrene Microplate,
V-bottom clear, polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3896
Corning
®
96 Well Clear Polystyrene Microplate,
V-bottom clear, polystyrene, pkg of Individually wrapped, sterile, lid: no
Expand
View Pricing
CLS3795
Corning
®
96 Well Clear Polystyrene Microplate,
round bottom clear, polystyrene, bag of 25, sterile, lid: no
Expand
View Pricing
CLS3628
Corning
®
96 Well Clear Polystyrene Microplate,
flat bottom clear, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing
CLS3360
Corning
®
96 Well Clear Polystyrene Microplate,
round bottom clear, Tissue Culture (TC)-treated surface, ampule of 25, sterile, lid: no
Expand
View Pricing
CLS3798
Corning
®
96 Well Clear Polystyrene Microplate,
round bottom clear, polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3898
Corning
®
96 Well Clear Polystyrene Microplate,
V-bottom clear, polystyrene, bag of 25, non-sterile, lid: no
Expand
View Pricing
CLS3358
Corning
®
96 Well Clear Polystyrene Microplate,
round bottom clear, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid
Expand
View Pricing